Skip to Content
Merck
All Photos(3)

Key Documents

I19209

Sigma-Aldrich

Isophthalic acid

99%

Synonym(s):

Benzene-1,3-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Beilstein:
1909332
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

autoignition temp.

1198 °F

mp

341-343 °C (lit.)

SMILES string

OC(=O)c1cccc(c1)C(O)=O

InChI

1S/C8H6O4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

QQVIHTHCMHWDBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isophthalic acid can be used as a reactant to prepare:
  • Poly(m-phenyleneisophthalamide) by polycondensation with m-phenylenediamine via thermal solid-phase polymerization reaction.[1]

It can also be used as a ligand to synthesize:
  • Metal coordination polymers by hydrothermal and sonochemical process.[2]
  • Isophthalic acid-zirconium(IV) nanocomposite, which is used as a precursor to prepare crystalline tetragonal ZrO2 via thermal decomposition method.[3]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue-Xiu Si et al.
International journal of biological macromolecules, 48(4), 700-704 (2011-03-05)
Using inhibition kinetics and computational simulation, we studied the reversible inhibition of tyrosinase by isophthalic acid (IPA). IPA inhibited tyrosinase in a complex manner with K(i)=17.8 ± 1.8mM. Measurements of intrinsic and ANS-binding fluorescence showed that IPA induced no changes
Suman Kalyan Pal et al.
Dalton transactions (Cambridge, England : 2003), (45)(45), 10021-10031 (2009-11-12)
Pyrene chromophores carrying different rigid rod spacer groups (ethynylene, ethynylene-phenylene-ethynylene, and ethynylene-bicyclo[2.2.2]octylene-ethynylene) and bound to TiO2 nanostructured materials via an isophthalic acid (Ipa) anchor group have been investigated using quantum chemical calculations in order to elucidate structural and electronic properties
Sonochemical synthesis of two nano-sized nickel (II) coordination polymers derived from flexible bis (benzimidazole) and isophthalic acid ligands
Li Jun-Xuan, et al.
Polyhedron, 160, 92-100 (2019)
Bo Xiao et al.
Nature chemistry, 1(4), 289-294 (2009-07-01)
Metal organic frameworks (MOFs) are among the most exciting materials discovered recently, attracting particular attention for their gas-adsorption and -storage properties. Certain MOFs show considerable structural flexibility in response to various stimuli. Although there are several examples of 'breathing' MOFs
Jörg Lincke et al.
Inorganic chemistry, 51(14), 7579-7586 (2012-07-04)
An isomorphous series of 10 microporous copper-based metal-organic frameworks (MOFs) with the general formulas (∞)(3)[{Cu(3)(μ(3)-OH)(X)}(4){Cu(2)(H(2)O)(2)}(3)(H-R-trz-ia)(12)] (R = H, CH(3), Ph; X(2-) = SO(4)(2-), SeO(4)(2-), 2 NO(3)(2-) (1-8)) and (∞)(3)[{Cu(3)(μ(3)-OH)(X)}(8){Cu(2)(H(2)O)(2)}(6)(H-3py-trz-ia)(24)Cu(6)]X(3) (R = 3py; X(2-) = SO(4)(2-), SeO(4)(2-) (9, 10)) is presented

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service