Merck
  • Shape Effect on Particle-Lipid Bilayer Membrane Association, Cellular Uptake, and Cytotoxicity.
Shape Effect on Particle-Lipid Bilayer Membrane Association, Cellular Uptake, and Cytotoxicity.
ACS applied materials & interfaces (2015-10-16)
Thapakorn Tree-Udom, Jiraporn Seemork, Kazuki Shigyou, Tsutomu Hamada, Naunpun Sangphech, Tanapat Palaga, Numpon Insin, Porntip Pan-In, Supason Wanichwecharungruang
ABSTRACT
Although computer simulation and cell culture experiments have shown that elongated spherical particles can be taken up into cells more efficiently than spherical particles, experimental investigation on effects of these different shapes over the particle-membrane association has never been reported. Therefore, whether the higher cellular uptake of an elongated spherical particles is a result of a better particle-membrane association as suggested by some calculation works or a consequence of its influence on other cellular trans-membrane components involved in particle translocation process, cannot be concluded. Here, we study the effect of particle shape on the particle-membrane interaction by monitoring the association between particles of various shapes and lipid bilayer membrane of artificial cell-sized liposomes. Among the three shaped lanthanide-doped NaYF4 particles, all with high shape purity and uniformity, similar crystal phase, and surface chemistry, the elongated spherical particle shows the highest level of membrane association, followed by the spherical particle with a similar radius, and the hexagonal prism-shaped particle, respectively. The free energy of membrane curvature calculated based on a membrane indentation induced by a particle association indicates that among the three particle shapes, the elongated spherical particle give the most stable membrane curvature. The elongated spherical particles show the highest cellular uptake into cytosol of human melanoma (A-375) and human liver carcinoma (HepG2) cells when observed through a confocal laser scanning fluorescence microscope. Quantitative study using flow cytometry also gives the same result. The elongated spherical particles also possess the highest cytotoxicity in A-375 and normal skin (WI-38) cell lines, comparing to the other two shaped particles.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
6-Aminofluorescein
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
6-Aminofluorescein, BioReagent, suitable for fluorescence, ~95% (HPLC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
SAFC
HEPES
Sigma-Aldrich
Ammonium fluoride, ACS reagent, ≥98.0%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)