Skip to Content
Merck
All Photos(3)

Key Documents

S8045

Sigma-Aldrich

Sodium hydroxide

BioXtra, ≥98% (acidimetric), pellets (anhydrous)

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352316
PubChem Substance ID:
NACRES:
NA.21

vapor density

>1 (vs air)

Quality Level

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

product line

BioXtra

Assay

≥98% (acidimetric)

form

pellets (anhydrous)

technique(s)

cell culture | mammalian: suitable

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

mp

318 °C (lit.)

solubility

H2O: 1 M, clear, colorless

density

2.13 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.02%
Mg: ≤0.0005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium hydroxide has been used:
  • to adjust pH of solutions
  • in the process of scraping cells from culture plate
  • for the preparation of formaldehyde solution
  • for the sanitation of chromatography column

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification of antibodies using the synthetic affinity ligand absorbent MAbsorbent A2P.
Chhatre S et al.
Nature Protocols, 2, 1763-1763 (2007)
Determination of the CYP1A-inducing potential of single substances, mixtures and extracts of samples in the micro-EROD assay with H4IIE cells.
Schiwy A, et al.
Nature Protocols, 10, 1728-1728 (2015)
Cytology and Microscopy: Immunolocalization of Covalently Modified Histone Marks on Barley Mitotic Chromosomes.
Colas I et al.
Bio-protocol null
Detergent resistance as a tool in membrane research.
Lingwood D and Simons K
Nature Protocols, 2, 2159-2159 (2007)
Horacio Cardenas et al.
Epigenetics, 9(11), 1461-1472 (2014-12-04)
A key step in the process of metastasis is the epithelial-to-mesenchymal transition (EMT). We hypothesized that epigenetic mechanisms play a key role in EMT and to test this hypothesis we analyzed global and gene-specific changes in DNA methylation during TGF-β-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service