Skip to Content
Merck
All Photos(5)

Documents

P2256

Sigma-Aldrich

Sodium pyruvate

ReagentPlus®, ≥99%

Synonym(s):

α-Ketopropionic acid sodium salt, 2-Oxopropanoic acid sodium salt, Pyruvic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOONa
CAS Number:
Molecular Weight:
110.04
Beilstein:
3568341
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

powder

loss

≤0.5% loss on drying

mp

>300 °C (lit.)

solubility

H2O: 100 mg/mL, clear, colorless to faintly yellow

cation traces

heavy metals (as Pb): ≤20 ppm

storage temp.

2-8°C

SMILES string

[Na+].CC(=O)C([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1

InChI key

DAEPDZWVDSPTHF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium pyruvate (SP) is a simplest keto-acid. It crystallizes as very thin plates and belongs to the monoclinic system. Its utility in Knoevenagel condensation carried out in an aqueous medium has been examined. The standard molar enthalpy of formation and molar enthalpy of dissolution of SP at infinite dilution have been obtained. Its efficacy in treating mitochondrial DNA depletion syndromes has been investigated.

Application

Sodium pyruvate has been used:
  • To prepare Bull Tyrode′s albumin-lactate-pyruvate diluent.
  • In pyruvate tolerance test.
  • To prepare oligodendrocyte culture medium.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donald A Morgan et al.
Diabetes, 64(6), 1976-1987 (2015-01-22)
Melanocortin 4 receptor (MC4R) signaling mediates diverse physiological functions, including energy balance, glucose homeostasis, and autonomic activity. Although the lateral hypothalamic area (LHA) is known to express MC4Rs and to receive input from leptin-responsive arcuate proopiomelanocortin neurons, the physiological functions
The crystal structure of sodium pyruvate.
Tavale SS, et al.
Acta Crystallographica, 14(12), 1281-1286 (1961)
Pyruvate therapy for mitochondrial DNA depletion syndrome.
Saito K, et al.
Biochim. Biophys. Acta Gen. Subj., 1820(5), 632-636 (2012)
"On Water": Rapid Knoevenagel Condensation Using Sodium Pyruvate.
Heravi MM, et al.
Letters in Organic Chemistry, 3(4), 297-299 (2006)
Crystal structure and thermochemical properties of a novel coordination compound sodium pyruvate C3H3O3Na(s).
Gao ZF, et al.
The Journal of Chemical Thermodynamics, 78, 189-196 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service