Skip to Content
Merck
All Photos(3)

Documents

P3662

Sigma-Aldrich

Sodium pyruvate

Hybri-Max, powder, suitable for hybridoma

Synonym(s):

α-Ketopropionic acid sodium salt, 2-Oxopropanoic acid sodium salt, Pyruvic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOONa
CAS Number:
Molecular Weight:
110.04
Beilstein:
3568341
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

grade

Hybri-Max

Assay

≥99%

form

powder

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

mp

>300 °C (lit.)

solubility

H2O: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].CC(=O)C([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1

InChI key

DAEPDZWVDSPTHF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium pyruvate has been used as a component in maturation medium for oocyte evaluation.
Sodium pyruvate is used by cells as an easily accessible carbohydrate source. Additionally, it is involved with amino acid metabolism and initiates the Kreb′s cycle. The 100 mM solution should be diluted 1:100 for most cell culture applications.

Biochem/physiol Actions

Sodium pyruvate is known to be an effective therapeutic target for type II citrullinaemia which is characterized by hyperammonaemia. In vivo studies proves that sodium pyruvate has protective function against hemorrhagic shock by preventing lipid peroxidation, NAD+ reduction and cleavage of poly(ADP-ribose) polymerases. Sodium pyruvate is also known to possess anti-inflammatory action and might show improvement in chronic lung disorder.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essence of Anesthesia Practice: Expert Consult-Online and Print, 685-685 (2010)
Treatment of a citrin-deficient patient at the early stage of adult-onset type II citrullinaemia with arginine and sodium pyruvate
Mutoh K, et al.
Journal of inherited metabolic disease, 31(2), 343-347 (2008)
Developmental competence and mRNA expression of preimplantation in vitro-produced embryos from prepubertal and postpubertal cattle and their relationship with apoptosis after intraovarian administration of IGF-1
Zaraza J, et al.
Theriogenology, 74(1), 75-89 (2010)
Metabolic and histologic effects of sodium pyruvate treatment in the rat after cortical contusion injury
Fukushima M, et al.
Journal of Neurotrauma, 26(7), 1095-1110 (2009)
Ovum pick up and in vitro embryo production in cows superstimulated with an individually adapted superstimulation protocol
De R, et al.
Animal Reproduction Science, 86(1-2), 13-25 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service