Merck
  • Wide-range screening of psychoactive substances by FIA-HRMS: identification strategies.
Wide-range screening of psychoactive substances by FIA-HRMS: identification strategies.
Analytical and bioanalytical chemistry (2015-04-12)
Élida Alechaga, Encarnación Moyano, Maria Teresa Galceran
ABSTRACT
Recreational drugs (illicit drugs, human and veterinary medicines, legal highs, etc.) often contain lacing agents and adulterants which are not related to the main active ingredient. Serious side effects and even the death of the consumer have been related to the consumption of mixtures of psychoactive substances and/or adulterants, so it is important to know the actual composition of recreational drugs. In this work, a method based on flow injection analysis (FIA) coupled with high-resolution mass spectrometry (HRMS) is proposed for the fast identification of psychoactive substances in recreational drugs and legal highs. The FIA and HRMS working conditions were optimized in order to detect a wide range of psychoactive compounds. As most of the psychoactive substances are acid-base compounds, methanol-0.1 % aqueous formic acid (1:1 v/v) as a carrier solvent and electrospray in both positive ion mode and negative ion mode were used. Two data acquisition modes, full scan at high mass resolution (HRMS) and data-dependent tandem mass spectrometry (ddMS/HRMS) with a quadrupole-Orbitrap mass analyzer were used, resulting in sufficient selectivity for identification of the components of the samples. A custom-made database containing over 450 substances, including psychoactive compounds and common adulterants, was built to perform a high-throughput target and suspect screening. Moreover, online accurate mass databases and mass fragmenter software were used to identify unknowns. Some examples, selected among the analyzed samples of recreational drugs and legal highs using the FIA-HRMS(ddMS/HRMS) method developed, are discussed to illustrate the screening strategy used in this study. The results showed that many of the analyzed samples were adulterated, and in some cases the sample composition did not match that of the supposed marketed substance.
Product Description
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
(−)-Levamisole hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile