Skip to Content
Merck
All Photos(2)

Key Documents

L1026

Sigma-Aldrich

Lidocaine

analytical standard

Synonym(s):

2-Diethylamino-N-(2,6-dimethylphenyl)acetamide, Lignocaine, Xylocaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O
CAS Number:
Molecular Weight:
234.34
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

grade

analytical standard

form

solid

packaging

vial of 200 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% Total Organic Impurities (Lidocaine related compound H ≤0.10%


2,6-Dimethylaniline ≤0.01%


Any unspecified impurity ≤0.10%


Total impurities ≤0.5%)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

SMILES string

CCN(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C14H22N2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)

InChI key

NNJVILVZKWQKPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lidocaine is an amide anesthetic, which is used to treat ventricular arrhythmias. It lowers cardiac automaticity and myocardial conduction. Lidocaine inhibits the generation and conduction of nerve impulses. It has an anticonvulsant effect.

Application

Lidocaine has been used to examine its effects in inhibiting lymph flow and protection against envenomation. It has also been used to assess its efficacy as an anti-tussive agent.

Biochem/physiol Actions

Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.
Na+ channel blocker; class IB antiarrhythmic.

Packaging

Supplied in amber screw-cap vials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of within-group designs to test anti-tussive drugs in conscious guinea-pigs
Venkatasamy R, et al.
Journal of Pharmacological and Toxicological Methods, 61(2), 157-162 (2010)
Pharmacological approaches that slow lymphatic flow as a snakebite first aid
van Helden DF, et al.
PLoS Neglected Tropical Diseases, 8(2), e2722-e2722 (2014)
Key Issues in Plastic and Cosmetic Surgery (1999)
Effect of lidocaine on ventricular arrhythmias in patients with coronary heart disease
Gianelly R, et al.
The New England Journal of Medicine, 277(23), 1215-1219 (1967)
Anesthesiology Keywords Review (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service