Skip to Content
Merck
All Photos(2)

Key Documents

C7731

Sigma-Aldrich

Caffeine

meets USP testing specifications, anhydrous

Synonym(s):

1,3,7-Trimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
Beilstein:
17705
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Agency:
USP/NF
meets USP testing specifications
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Agency

USP/NF
meets USP testing specifications

form

crystalline
powder

mp

234-236.5 °C (lit.)

solubility

H2O: soluble 18.7 g/L at 16 °C

SMILES string

CN1C(=O)N(C)c2ncn(C)c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Caffeine is a commonly consumed pharmacologically active substance. It is a member of the class of methylxanthines.[1] It is a natural alkaloid found in coffee, cocoa and other plants. Caffeine might be linked to osteoporosis, cardiovascular risk, arrhythmia and gastrointestinal disturbances.[2] Caffeine is used to treat orthostatic hypotension,[3] bronchopulmonary dysplasia and apnea of prematurity (AOP).[4]

Application

Caffeine has been used:
  • to functionally disable Ca2+ removal via sarco/endoplasmic reticulum Ca2+ ATPase (SERCA)[5]
  • to determine its effects in overcoming excitation-contraction-coupling failure[6]
  • to compare its role as a stimulant with guarana[7]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

There and back again: Iterating between population-based modeling and experiments reveals surprising regulation of calcium transients in rat cardiac myocytes
Devenyi RA and Sobie EA
Journal of Molecular and Cellular Cardiology, 96, 38-48 (2016)
Orthostatic hypotension in the elderly: diagnosis and treatment
Gupta V and Lipsitz LA
The American Journal of Medicine, 120(10), 841-847 (2007)
Metabolic effects of spices, teas, and caffeine
Westerterp-Plantenga M, et al.
Physiology & Behavior, 89(1), 85-91 (2006)
Ryan A Devenyi et al.
Journal of molecular and cellular cardiology, 96, 38-48 (2015-08-04)
While many ion channels and transporters involved in cardiac cellular physiology have been identified and described, the relative importance of each in determining emergent cellular behaviors remains unclear. Here we address this issue with a novel approach that combines population-based
Apnea of prematurity: from cause to treatment
Zhao J, et al.
European Journal of Pediatrics, 170(9), 1097-1105 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service