Skip to Content
Merck
All Photos(3)

Key Documents

E1501

Sigma-Aldrich

Benzocaine

≥99% (HPLC)

Synonym(s):

4-Aminobenzoic acid ethyl ester, Ethyl 4-aminobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Beilstein:
638434
EC Number:
MDL number:
UNSPSC Code:
51271900
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (HPLC)

form

powder

solubility

ethanol: soluble 50 mg/mL

SMILES string

CCOC(=O)c1ccc(N)cc1

InChI

1S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2,10H2,1H3

InChI key

BLFLLBZGZJTVJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzocaine is a topical anesthetic. It functions as an appetite suppressant and smoke inhibitor. Benzocaine is used to treat pruritus, burns and cold. It is associated with allergic contact dermatitis.

Application

Benzocaine has been used as an anesthetic in cerebrospinal fluid (CSF) flow analysis. It also has been used as an anesthetic in fishes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ciliogenesis and cerebrospinal fluid flow in the developing Xenopus brain are regulated by foxj1
Hagenlocher C, et al.
Cilia, 2(1), 12-12 (2013)
Fisher's Contact Dermatitis (2008)
Guillaume Castel et al.
Journal of virology, 83(20), 10808-10820 (2009-08-27)
We wanted to develop a therapeutic approach against rabies disease by targeting the lyssavirus transcription/replication complex. Because this complex (nucleoprotein N-RNA template processed by the L polymerase and its cofactor, the phosphoprotein P) is similar to that of other negative-strand
Consequences of acute stress and cortisol manipulation on the physiology, behavior, and reproductive outcome of female Pacific salmon on spawning grounds
McConnachie SH, et al.
Hormones and Behavior, 62(1), 67-76 (2012)
Early individual electronic identification of sea bass using RFID microtags: A first example of early phenotyping of sex-related growth
Ferrari S, et al.
Aquaculture (Amsterdam, Netherlands), 426, 165-171 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service