Skip to Content
Merck
All Photos(1)

Key Documents

L-018

Supelco

Lidocaine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O
CAS Number:
Molecular Weight:
234.34
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCN(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C14H22N2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)

InChI key

NNJVILVZKWQKPM-UHFFFAOYSA-N

General description

Lidocaine is a local anesthetic and antiarrhythmic drug. This analytical reference standard is suitable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis. Lidocaine is often added to cocaine as a diluent.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Soledad Cepeda et al.
The Cochrane database of systematic reviews, (12)(12), CD006581-CD006581 (2010-12-15)
Lidocaine administration produces pain due to its acidic pH. The objective of this review was to determine if adjusting the pH of lidocaine had any effect on pain resulting from non-intravascular injections in adults and children. We tested the hypothesis
Bettina F Cuneo et al.
Circulation, 128(20), 2183-2191 (2013-11-13)
The electrophysiology of long QT syndrome (LQTS) in utero is virtually unstudied. Our goal here was to evaluate the efficacy of fetal magnetocardiography (fMCG) for diagnosis and prognosis of fetuses at risk of LQTS. We reviewed the pre/postnatal medical records
Jason Wasiak et al.
The Cochrane database of systematic reviews, 6(6), CD005622-CD005622 (2012-06-15)
This is an update of the review on 'Lidocaine for pain relief in burn injured patients' first published in Issue 3, 2007. Pain is a major issue for patients suffering from many different types of wounds, in particular those with
Gérard Mick et al.
Current medical research and opinion, 28(6), 937-951 (2012-05-04)
The topical 5% lidocaine medicated plaster is recommended as first-line treatment for localized peripheral neuropathic pain. In order to provide an overview of the efficacy and safety of the lidocaine plaster in the treatment of different neuropathic pain conditions, all
Frederick A Matsen et al.
The Journal of bone and joint surgery. American volume, 95(12), 1126-1134 (2013-06-21)
Glenohumeral chondrolysis is the irreversible destruction of previously normal articular cartilage, occurring most commonly after shoulder surgery in young individuals. The reported incidence of this complication has risen rapidly since the early 2000s. As chondrolysis cannot be reversed, its occurrence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service