Skip to Content
Merck
All Photos(4)

Key Documents

271004

Sigma-Aldrich

Acetonitrile

Acetonitrile
2 of 2 reviewers received a sample product or took part in a promotion

99.8%, anhydrous, suitable for solid phase extraction (SPE)

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
12191502
eCl@ss:
39031501
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
technique(s):
solid phase extraction (SPE): suitable
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Acetonitrile, anhydrous, 99.8%

grade

anhydrous

Quality Level

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

99.8%

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

solid phase extraction (SPE): suitable

impurities

<0.001% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

color

colorless

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

solubility

water: soluble (completely)

density

0.786 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile, an aliphatic nitrile, is widely used as an organic solvent and intermediate in organic syntheses.[1] It is transparent to UV-visible light, which makes it highly applicable in spectrophotometric and fluorimetric techniques. MeCN is utilized as a mobile phase component in many chromatographic techniques, due to its low viscosity, high elution strength and miscibility in water. It also plays a major role as an extractant medium in liquid-liquid extraction, solid-phase extraction or microextraction.[2]

Application

Acetonitrile may be used as a solvent to prepare:
  • 1,2-Azidoalcohols and 1,2-azidoamines via cerium(III) chloride assisted ring opening of epoxides and aziridines by sodium azide.[3]
  • Cyano-bearing indolinones by oxidative arylalkylation of olefins in the presence of palladium catalyst.[4]

It may also be used as a reactant to synthesize:
  • Bis (diphenylphosphino) acetonitrile by reacting with n-butyllithium and then with chlorodiphenylphosphine.[5]
  • β-Acetamido ketones via coupling reaction with ketones or ketoesters and aldehydes in the presence of cobalt(II) chloride.[6]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Bis (diphenylphosphino) acetonitrile: Synthesis, ligand properties and application in catalytic carbon?carbon coupling
Braun L, et al.
Dalton Transactions, 14, 1409-1415 (2007)
Cobalt-catalysed three-component coupling involving ketones or ketoesters, aldehydes and acetonitrile: a novel one-pot synthesis of ?-acetamido ketones
Madhavaa Reddy M.
Journal of the Chemical Society. Chemical Communications, 6, 713-714 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 56-56 (1994)
Cerium (III) chloride promoted highly regioselective ring opening of epoxides and aziridines using NaN3 in acetonitrile: a facile synthesis of 1,2-azidoalcohols and 1,2-azidoamines.
Sabitha G, et al.
Organic Letters, 4(3), 343-345 (2002)
Palladium-Catalyzed Oxidative Arylalkylation of Activated Alkenes: Dual C-H Bond Cleavage of an Arene and Acetonitrile.
Wu T, et al.
Angewandte Chemie (International Edition in English), 50(52), 12578-12581 (2011)

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Hi! By which method is the acetonitrile dried? Is molecular sieve used? Thanks!

    1 answer
    1. Unfortunately, the manufacturing process of this product is proprietary. If necessary, molecular sieves can help to further dry the product before use.

      Helpful?

  4. Hey, my name is Mohammad. We tried to develop sensor ro detect heavy metals. And I fond some peaks in the solution when I added acn 99.8% and I want to know if there is heavy metals and which metals?

    1 answer
    1. Unfortunately, this product is not tested for heavy metals. Should this be a critical issue for the specific application, see product 900667. This product is tested for trace metals and may be a better option. See the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/900667-BULKSHBR1056.pdf

      Helpful?

Reviews

2 of 2 reviewers received a sample product or took part in a promotion
1–2 of 2 Reviews  

Active Filters

  1. Cambridge
    • Reviews 7
    • Votes 15
    5 out of 5 stars.

    Anhydrous acetonitrile

    Good quality acetonitrile. Anhydrous status has been maintained.

    Helpful?

  2. Sydney
    • Review 1
    • Votes 2
    5 out of 5 stars.

    Works well

    We have used Sigma Aldrich's anhydrous acetonitrile (271004) for the synthesis of synthetic RNA and it works well. We have recommended it to other researchers as it saves time and is safer than distilling anhydrous acetonitrile.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service