Merck
  • Conformation and activity of lipase B from Candida antarctica in bicontinuous microemulsions.
Conformation and activity of lipase B from Candida antarctica in bicontinuous microemulsions.
Colloids and surfaces. B, Biointerfaces (2015-05-15)
Mireia Subinya, Anne K Steudle, Tomasz P Jurkowski, Cosima Stubenrauch
ABSTRACT
The paper at hand deals with the influence of the pH-value on the conformation and activity of the lipase B from Candida antarctica (CalB) which is incorporated in a bicontinuous microemulsion. The microemulsion used for this purpose consists of water/NaCl, n-octane, and the non-ionic surfactant penthaethylene glycol monodecylether (C10E5). The conformational study clearly shows (1) that CalB molecules are partitioned between the interfacial monolayer and the water domains and (2) that the pH-value of the microemulsion's water domains strongly influences the conformation of CalB at the interfacial monolayer. From these observations we conclude that there is a continuous exchange between the CalB molecules, which are located at the interfacial monolayer and those which are located in the water domains of the microemulsion. This exchange strongly influences the CalB conformation in both regions. In addition to the conformation, we also studied the catalytic activity of CalB. The catalytic measurements revealed a bell-shaped dependence between the CalB activity and the pH-value. The maximum catalytic activity of CalB in bicontinuous microemulsions was observed at pH=5.5. At this pH we observed the highest amount of α-helix conformation of the CalB molecules that are located at the interfacial monolayer, which, in turn, allows connecting the activity with the conformation.
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Pentaethylene glycol monodecyl ether, BioXtra, ≥97.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Pentaethylene glycol, 98%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)