Skip to Content
Merck
All Photos(1)

Documents

335754

Sigma-Aldrich

Pentaethylene glycol

98%, average MN 250

Synonym(s):

Polyethylene glycol, 3,6,9,12-Tetraoxatetradecane-1,14-diol, Pentaglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)5H
CAS Number:
Molecular Weight:
238.28
Beilstein:
1635593
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

product name

Pentaethylene glycol, 98%

Assay

98%

form

liquid

mol wt

average Mn 250

refractive index

n20/D 1.462 (lit.)

bp

184 °C/2 mmHg (lit.)

density

1.126 g/mL at 25 °C (lit.)

Ω-end

hydroxyl

α-end

hydroxyl

SMILES string

OCCOCCOCCOCCOCCO

InChI

1S/C10H22O6/c11-1-3-13-5-7-15-9-10-16-8-6-14-4-2-12/h11-12H,1-10H2

InChI key

JLFNLZLINWHATN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Application of Prussian Blue in Electrochemical and Optical Sensing of Free Chlorine.: This study discusses the use of Prussian Blue on fluorine-doped tin oxide coated glass slides for electrochemical and optical sensing applications, specifically targeting free chlorine detection, highlighting its high sensitivity and stability (Valiūnienė et al., 2022).

  • Evaluation of a photoelectrochemical platform based on strontium titanate, sulfur doped carbon nitride and palladium nanoparticles for detection of SARS-CoV-2 spike glycoprotein S1.: The article evaluates the integration of fluorine-doped tin oxide in a complex photoelectrochemical cell designed for the detection of COVID-19′s spike protein, underscoring its potential in rapid diagnostic applications (Botelho et al., 2022).

  • Immunodiagnostic of leprosy exploiting a photoelectrochemical platform based on a recombinant peptide mimetic of a Mycobacterium leprae antigen.: This research highlights a novel use of fluorine-doped tin oxide slides in a photoelectrochemical setup for the sensitive and specific immunodiagnosis of leprosy, showcasing the material′s application in medical diagnostics (Yotsumoto Neto et al., 2019).

  • Fast and reliable BIA/amperometric quantification of acetylcysteine using a nanostructured double hydroxide sensor.: This paper presents a method utilizing fluorine-doped tin oxide for the rapid and reliable electrochemical detection of acetylcysteine, demonstrating the coating′s effectiveness in enhancing electron transfer and sensitivity in biosensing applications (Correa et al., 2018).

  • Nanostructured TiO2 Films Attached CdSe QDs Toward Enhanced Photoelectrochemical Performance.: Discusses the role of fluorine-doped tin oxide substrates in improving the photoelectrochemical properties of TiO2 films combined with quantum dots, which could advance the development of more efficient solar cells and sensors (Du et al., 2016).


Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Shi et al.
Nature communications, 10(1), 2223-2223 (2019-05-22)
Mammalian cells are different from plant and microbial cells, having no exterior cell walls for protection. Environmental assaults can easily damage or destroy mammalian cells. Thus, the ability to develop a biomimetic cell wall (BCW) on their plasma membrane as
Steven P D Harborne et al.
Scientific reports, 7, 45383-45383 (2017-03-30)
Mitochondrial ATP-Mg/Pi carriers import adenine nucleotides into the mitochondrial matrix and export phosphate to the cytosol. They are calcium-regulated to control the size of the matrix adenine nucleotide pool in response to cellular energetic demands. They consist of three domains:
Alexis Hofherr et al.
PLoS biology, 16(8), e2005651-e2005651 (2018-08-07)
Cilia are organelles specialized in movement and signal transduction. The ciliary transient receptor potential ion channel polycystin-2 (TRPP2) controls elementary cilia-mediated physiological functions ranging from male fertility and kidney development to left-right patterning. However, the molecular components translating TRPP2 channel-mediated
Stephany Jaiquel Baron et al.
Theranostics, 11(11), 5077-5091 (2021-04-17)
An increasing number of commonly prescribed drugs are known to interfere with mitochondrial function, causing cellular toxicity, but the underlying mechanisms are largely unknown. Although often not considered, mitochondrial transport proteins form a significant class of potential mitochondrial off-targets. So
Chelsea L Price et al.
The journal of physical chemistry. A, 124(9), 1811-1820 (2020-02-06)
Single-particle trapping is an effective strategy to explore the physical and optical properties of aerosol with high precision. Laser-based methods are commonly used to probe the size, optical properties, and composition of nonlight-absorbing droplets in optical and electrodynamic traps. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service