Skip to Content
Merck
All Photos(1)

Documents

S6546

Sigma-Aldrich

Sodium chloride solution

5 M

Synonym(s):

Halite, NaCl, sodium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25

sterility

0.2 μm filtered

Quality Level

concentration

5 M

density

1.190 g/cm3

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium chloride (NaCl), common salt also known as halite, is ubiquitous in occurrence and has many vital applications. It is a white crystalline solid, fairly soluble in water. NaCl is a flavouring agent in the food industry and used for texturing bread and baked foods. NaCl is an essential salt for normal body functions in humans and animals. NaCl is essential for maintaining plasma osmolality and extracellular fluid volume of the body. Excess of NaCl leads to elevated blood pressure which has implications on cardiovascular and renal diseases. NaCl is a basic component in the microbiological medium, specifically for culturing of halophiles. Sterile NaCl is a constituent of eye drop solutions.

Application

Sodium chloride has been used:
  • as a supplement in serum free media for CA3 pyramidal neuron layer preparation
  • in the preparation of oligonucleotides (ODNs) annealing buffer
  • in the preparation of phosphate buffered saline and in phosphatase methylesterase-1 (PME-1) inhibition and counterscreen assay buffers

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Actions and Uses of Ophthalmic Drugs: A Textbook for Students and Practitioners (2013)
Probe Reports from the NIH Molecular Libraries Program (2010)
Dietary Reference Intakes for Water, Potassium, Sodium, Chloride, and Sulfate (2005)
Simultaneous reprogramming and gene editing of human fibroblasts
Howden SE, et al.
Nature Protocols, 13(5), 875-875 (2018)
Salt, chlor-alkali and related heavy chemicals
Kent and Riegel's Handbook of Industrial Chemistry and Biotechnology (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service