Skip to Content
Merck
All Photos(2)

Key Documents

657042

Sigma-Aldrich

Octane

electronic grade, ≥99.999% metals basis, ≥99% (CP)

Synonym(s):

n-Octane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696875
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
grade:
electronic grade
bp:
125-127 °C (lit.)
vapor pressure:
11 mmHg ( 20 °C)

grade

electronic grade

vapor density

3.9 (vs air)

vapor pressure

11 mmHg ( 20 °C)

Assay

≥99% (CP)
≥99.999% metals basis

form

liquid

autoignition temp.

428 °F

expl. lim.

6.5 %

refractive index

n20/D 1.398 (lit.)

bp

125-127 °C (lit.)

mp

−57 °C (lit.)

density

0.703 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC

InChI

1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3

InChI key

TVMXDCGIABBOFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela J Sueldo et al.
The New phytologist, 208(1), 210-223 (2015-05-27)
Plant nucleotide-binding, leucine-rich repeat (NB-LRR) proteins confer immunity to pathogens possessing the corresponding avirulence proteins. Activation of NB-LRR proteins is often associated with induction of the hypersensitive response (HR), a form of programmed cell death. NRC1 (NB-LRR Required for HR-Associated
Xian-Long Du et al.
ChemSusChem, 8(20), 3489-3496 (2015-09-15)
The N-methylation of amines with CO2 and H2 is an important step in the synthesis of bioactive compounds and chemical intermediates. The first heterogeneous Au catalyst is reported for this methylation reaction with good to excellent yields. The average turnover
Mariela Ivanova et al.
Electrophoresis, 24(6), 992-998 (2003-03-27)
Microemulsion electrokinetic chromatography was applied for the separation of levetiracetam from other antiepileptic drugs (primidone, phenobarbital, phenytoin, lamotrigine and carbamazepine) that are potentially coadministered in therapy of patients. The influence of the composition of the microemulsion system (with sodium dodecyl
Ben Li et al.
Journal of hazardous materials, 211-212, 296-303 (2011-11-08)
The sulfur in gasoline will convert to SO(2) after combustion under high temperature, which adversely affects human health and the environment. Membrane technique in particular pervaporation offers a number of potential advantages over conventional FCC gasoline desulfurization processes. The present
Rama Krishna Gudiminchi et al.
Applied microbiology and biotechnology, 96(6), 1507-1516 (2012-03-14)
CYP153A6 is a well-studied terminal alkane hydroxylase which has previously been expressed in Pseudomonas putida and Escherichia coli by using the pCom8 plasmid. In this study, CYP153A6 was successfully expressed in E. coli BL21(DE3) by cloning the complete operon from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service