Skip to Content
Merck
All Photos(4)

Key Documents

M3671

Sigma-Aldrich

MES

low moisture content, ≥99% (titration)

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid, 4-Morpholineethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4S
CAS Number:
Molecular Weight:
195.24
Beilstein:
141319
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (titration)

form

powder

storage condition

dry at room temperature

impurities

≤1.0% water

color

white

pH

2.5-4 (25 °C, 97.6 g/L)

useful pH range

5.5-6.7

pKa 

6.1

density

1.23 g/cm3 at 20—25 °C
1.27 g/cm3 at 20—25 °C

application(s)

clinical research
diagnostic assay manufacturing
life science and biopharma (
)
metabolomics (
)

SMILES string

OS(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S/c8-12(9,10)6-3-7-1-4-11-5-2-7/h1-6H2,(H,8,9,10)

InChI key

SXGZJKUKBWWHRA-UHFFFAOYSA-N

General description

MES Buffer, recognized as one of the "Good" buffers employed in biological and biochemical applications, plays a crucial role across various research domains. This zwitterionic and morpholinic buffer proves highly valuable due to its effective buffering capacity within the pH range of 5.5 to 6.7. Notably, MES Buffer does not engage in complex formation with the majority of metals commonly employed in environmental and biological studies. Its solubility in water is excellent, and it exhibits minimal lipid solubility, rendering it impermeable to membranes.

This versatile buffer may find widespread application in maintaining a stable environment in solutions dedicated to cell culture media, protein-based buffer formulations, and electrophoresis running buffers. Its significance extends to the purification processes of antibodies, peptides, proteins, blood components, and growth factors. MES Buffer stands out as an optimal choice for capillary electrochromatography, offering low ionic mobility. Notably, it serves as a safer alternative to potentially toxic agents like cacodylate and non-zwitterionic buffers, including citrate and malate. MES Buffer also proves valuable for fine-tuning the pH of growth media, stabilizing enzymatic solutions, and serving as a component of PAGE running buffer in diverse experimental settings.

Application

It has been utilized as a buffer bicarbonate‐containing culture medium
MES has been used as a component of the elution buffer used to elute plasma from α2-macroglobulin affinity column. It has been used as an activation buffer during antibody conjugation to microparticles. MES has been used in the functionalization of microchannels during the creation of microfluidic-integrated surface plasmon resonance (SPR) platform for pathogen detection.

Features and Benefits

  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Effective Buffering from pH 5.5-6.7 (25 °C) with a pKa of 6.1 (25 °C)
  • Highly soluble in water
  • Minimal metal ion binding
  • Less toxic to cells than other buffers such as Tris and phosphate
  • Stable in a wide pH range
  • Low UV absorptivity and Minimal reactivity

Quality

Low moisture

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impaired lipoprotein receptor-mediated peripheral binding of plasma amyloid-? is an early biomarker for mild cognitive impairment preceding Alzheimer's disease.
Sagare, Abhay P., et al.
Journal of Alzheimer'S Disease, 24 (1), 25-34 (2011)
Portable microfluidic integrated plasmonic platform for pathogen detection
Tokel, Onur, et al.
Scientific Reports, 2015, 9152-9152 null
Sensitive Mie scattering immunoagglutination assay of porcine reproductive and respiratory syndrome virus (PRRSV) from lung tissue samples in a microfluidic chip
Song, Jae-Young, et al.
J. Virol. Methods, 178, 31-38 (2011)
"(Un) suitability of the use of pH buffers in biological, biochemical and environmental studies and their interaction with metal ions-a review.
Ferreira, Carlos MH, et al.
Royal Society of Chemistry Advances, 5 (39), 30989-31003 (2015)
Sensitive Mie scattering immunoagglutination assay of porcine reproductive and respiratory syndrome virus (PRRSV) from lung tissue samples in a microfluidic chip.
Song JY, Lee CH, Choi EJ, Kim K, et al.
J. Virol. Methods, 178, 31-38 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service