Skip to Content
Merck
All Photos(2)

Documents

378860

Sigma-Aldrich

Sodium chloride

random crystals, optical grade, 99.9% trace metals basis

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

optical grade

vapor pressure

1.33 (1.00 mmHg) at 865 °C hPa

Assay

99.9% trace metals basis

form

random crystals

technique(s)

IR spectroscopy: suitable

impurities

≤1500.0 ppm Trace Metal Analysis

pH

7

bp

1465 °C/1 atm (lit.)

mp

801 °C (lit.)

solubility

ethanol: slightly soluble(lit.)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Francis Kim et al.
JAMA, 311(1), 45-52 (2013-11-19)
Hospital cooling improves outcome after cardiac arrest, but prehospital cooling immediately after return of spontaneous circulation may result in better outcomes. To determine whether prehospital cooling improves outcomes after resuscitation from cardiac arrest in patients with ventricular fibrillation (VF) and
Frédéric Lebrun-Julien et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(25), 8432-8448 (2014-06-21)
The mammalian target of rapamycin (mTOR) pathway integrates multiple signals and regulates crucial cell functions via the molecular complexes mTORC1 and mTORC2. These complexes are functionally dependent on their raptor (mTORC1) or rictor (mTORC2) subunits. mTOR has been associated with
Alexander W Wyatt et al.
Genome biology, 15(8), 426-426 (2014-08-27)
Genomic analyses of hundreds of prostate tumors have defined a diverse landscape of mutations and genome rearrangements, but the transcriptomic effect of this complexity is less well understood, particularly at the individual tumor level. We selected a cohort of 25
Gemma L Carvill et al.
Neurology, 82(14), 1245-1253 (2014-03-14)
To determine the genes underlying Dravet syndrome in patients who do not have an SCN1A mutation on routine testing. We performed whole-exome sequencing in 13 SCN1A-negative patients with Dravet syndrome and targeted resequencing in 67 additional patients to identify new
I Dzneladze et al.
Leukemia, 29(7), 1485-1495 (2015-03-05)
In this study, we investigated the role of inositol polyphosphate-4-phosphatase, type-II (INPP4B) in acute myeloid leukemia (AML). We observed that AML patients with high levels of INPP4B (INPP4B(high)) had poor response to induction therapy, shorter event-free survival and shorter overall

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service