Merck
  • Effects of cyano-substituents on the molecular packing structures of conjugated polymers for bulk-heterojunction solar cells.
Effects of cyano-substituents on the molecular packing structures of conjugated polymers for bulk-heterojunction solar cells.
ACS applied materials & interfaces (2014-08-26)
Hyojung Cha, Hyoung Nam Kim, Tae Kyu An, Moon Sung Kang, Soon-Ki Kwon, Yun-Hi Kim, Chan Eon Park
ABSTRACT
The molecular packing structures of two conjugated polymers based on alkoxy naphthalene, one with cyano-substituents and one without, have been investigated to determine the effects of electron-withdrawing cyano-groups on the performance of bulk-heterojunction solar cells. The substituted cyano-groups facilitate the self-assembly of the polymer chains, and the cyano-substituted polymer:PC71BM blend exhibits enhanced exciton dissociation to PC71BM. Moreover, the electron-withdrawing cyano-groups lower the highest occupied molecular orbital (HOMO) and lowest unoccupied molecular orbital (LUMO) levels of the conjugated polymer, which leads to a higher open circuit voltage (V(OC)) and a lower energy loss during electron transfer from the donor to the acceptor. A bulk-heterojunction device fabricated with the cyano-substituted polymer:PC71BM blend has a higher V(OC) (0.89 V), a higher fill factor (FF) (51.4%), and a lower short circuit current (J(SC)) (7.4 mA/cm(2)) than that of the noncyano-substituted polymer:PC71BM blend under AM 1.5G illumination with an intensity of 100 mW cm(-2). Thus, the cyano-substitution of conjugated polymers may be an effective strategy for optimizing the domain size and crystallinity of the polymer:PC71BM blend, and for increasing V(OC) by tuning the HOMO and LUMO energy levels of the conjugated polymer.
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
1,5-Dihydroxynaphthalene, 97%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate ChemBeads
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Phenylboronic acid, 95%
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Bromine, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
n-Butyllithium solution, 2.7 M in heptane
Sigma-Aldrich
Phenylboronic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
n-Butyllithium solution, 11.0 M in hexanes
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane