Skip to Content
Merck
All Photos(3)

Documents

S7795

Sigma-Aldrich

Sodium carbonate

BioXtra, ≥99.0%

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.21

product line

BioXtra

Quality Level

Assay

≥99.0%

form

powder

impurities

≤0.02% Phosphorus (P)
≤0.1% Insoluble matter

pH

11.2 (25 °C, 4 g/L)

mp

851 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.02%

cation traces

Al: ≤0.001%
Ca: ≤0.02%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carbonate is usually referred to as soda ash. Solvay process uses sodium chloride and limestone to produce sodium carbonate. Sodium carbonate enhances the pretreatment of rice straw and enhances delignification process. Sodium carbonate finds its application in the extraction of bioactive metabolites like epoxyditerpenoids and alkaloids and removes acidic compounds from the extract. Sodium carbonate induces the phase separation in salt induced phase separation extraction (SIPSE) method and enhances the plasma partitioning with acetonitrile.

Application

Sodium carbonate has been used:
  • to stop α-glucosidase enzyme activity using the extract of Terminalia species
  • as a component in carbonate/bicarbonate buffer for enzyme linked immunosorbent assay (ELISA)
  • as a component in the growth medium for Synechocystis strains

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Carbonate: A Versatile Material (2000)
ELISA to evaluate plasma anti-asparaginase IgG concentrations in patients with acute lymphoblastic leukemia
Wang B, et al.
Journal of Immunological Methods, 239(1-2), 75-83 (2000)
Analysis of Metals in Whole Cells, Thylakoids and Photosynthetic Protein Complexes in Synechocystis sp. PCC6803
Gandini C, et al.
Bio-protocol, 8(12), e2889-e2889 (2018)
alpha-glucosidase inhibitor activity of Terminalia species
Anam K, et al.
Int. J. Pharmacol., 5(9), 277-280 (2009)
Eduardo Isanta et al.
Water research, 80, 149-158 (2015-05-23)
Partial nitritation for a low-strength wastewater at low temperature was stably achieved in an aerobic granular reactor. A bench-scale granular sludge bioreactor was operated in continuous mode treating an influent of 70 mg N-NH4(+) L(-1) to mimic pretreated municipal nitrogenous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service