60377
Potassium hydroxide
tested according to Ph. Eur.
Synonym(s):
Kalii hydroxidum, Caustic potash
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
Agency
USP/NF
tested according to Ph. Eur.
Quality Level
vapor pressure
1 mmHg ( 719 °C)
Assay
85.00%
form
solid
mp
361 °C (lit.)
SMILES string
[OH-].[K+]
InChI
1S/K.H2O/h;1H2/q+1;/p-1
InChI key
KWYUFKZDYYNOTN-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Andreu Fabregat et al.
Steroids, 78(3), 327-336 (2012-12-25)
The occurrence of several polyunsaturated testosterone related compounds (including 4,6-androstadien-3,17-dione and 4,6-androstadien-17β-ol-3-one) in urine after alkaline treatment of the sample has been recently reported. Although several experiments seem to indicate that they are testosterone metabolites, their origin is still unknown.
Kristina Nyström et al.
PLoS pathogens, 7(8), e1002188-e1002188 (2011-09-09)
Rabbit Hemorrhagic disease virus (RHDV), a calicivirus of the Lagovirus genus, and responsible for rabbit hemorrhagic disease (RHD), kills rabbits between 48 to 72 hours post infection with mortality rates as high as 50-90%. Caliciviruses, including noroviruses and RHDV, have
Eduardo J M Paiva et al.
Ultrasonics sonochemistry, 20(3), 833-838 (2012-12-05)
Ethyl esters of babassu oil were synthesized by alkaline catalysis to make the green production of biodiesel feasible with simple methods and available technology. Babassu oil is a transparent, light yellow oil extracted from the seeds of the babassu palm
Hui Boon Teh et al.
Journal of chromatography. A, 1383, 112-120 (2015-02-05)
A new, highly sensitive and reliable two-dimensional matrix elimination ion chromatography (IC) method was developed for simultaneous detection of bromate, chlorite and five haloacetic acids. This method combined the conventional IC in first dimension with capillary IC in the second
Heng-guo Wang et al.
ChemSusChem, 6(1), 56-60 (2012-12-12)
Between the sheets: Sodium-ion batteries are an attractive, low-cost alternative to lithium-ion batteries. Nitrogen-doped porous carbon sheets are prepared by chemical activation of polypyrrole-functionalized graphene sheets. When using the sheets as anode material in sodium-ion batteries, their unique compositional and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service