Skip to Content
Merck
All Photos(4)

Documents

409286

Sigma-Aldrich

Sodium iodide

99.999% trace metals basis

Synonym(s):

Sodium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaI
CAS Number:
Molecular Weight:
149.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

Assay

99.999% trace metals basis

form

crystalline

impurities

≤15.0 ppm Trace Metal Analysis

mp

661 °C (lit.)

SMILES string

[Na+].[I-]

InChI

1S/HI.Na/h1H;/q;+1/p-1

InChI key

FVAUCKIRQBBSSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium iodide (NaI) is a water soluble ionic compound that consists of an equivalent addition of sodium as a cation (Na+) and iodide as an anion (I-) in crystal space. It is industrially prepared by reacting acidic iodides with sodium hydroxide (NaOH) to form NaI salts. It is a less toxic compound than iodine and can be stored and handled easily.

Application

NaI can be used as a precursor in the preparation of controlling agents for emulsion polymerization. It can also be used as an inorganic salt in the development of liquid electrolytes for the fabrication of dye sensitized solar cells (DSSCs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodine and iodine compounds
Lyday PA and Kaiho T
Ullmann's Encyclopedia of Industrial Chemistry, 1-13 (2000)
Gel polymer electrolyte based on poly (acrylonitrile-co-styrene) and a novel organic iodide salt for quasi-solid state dye-sensitized solar cell
Wu J, et al.
Electrochimica Acta, 51(20), 4243-4249 (2006)
Use of sodium iodide as the precursor to the control agent in ab initio emulsion polymerization
Tonnar J and Lacroix-Desmazes P
Angewandte Chemie (International Edition in English), 47(7), 1294-1297 (2008)
Shuzhong Zhang et al.
Frontiers in immunology, 11, 1871-1871 (2020-09-26)
Macrophage dysfunction is fundamentally related to altered immunity in cystic fibrosis (CF). How genetic deficits in the cystic fibrosis transmembrane conductance regulator (CFTR) lead to these defects remains unknown. Rapid advances in genomic editing such as the clustered regularly interspaced
Dafu Yu et al.
Hormones and behavior, 69, 106-115 (2015-01-28)
Thyroid hormone disorders have long been linked to depression, but the causal relationship between them remains controversial. To address this question, we established rat models of hypothyroidism using (131)iodine ((131)I) and hyperthyroidism using levothyroxine (LT4). Serum free thyroxine (FT4) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service