Skip to Content
Merck
All Photos(1)

Key Documents

470864

Sigma-Aldrich

Bromine

≥99.99% trace metals basis

Synonym(s):

Dibromine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Br2
CAS Number:
Molecular Weight:
159.81
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.14 (vs air)

Quality Level

vapor pressure

175 mmHg ( 20 °C)
671 mmHg ( 55 °C)

Assay

≥99.5% (by Na2S2O3, titration)
≥99.99% trace metals basis

resistivity

7.8E18 μΩ-cm, 20°C

impurities

≤0.001% I2
≤0.001% S compounds
<100 ppm total metallic impurities

evapn. residue

≤0.005%

bp

58.8 °C (lit.)

mp

−7.2 °C (lit.)

density

3.119 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.05%

cation traces

Ni: ≤5 ppm
heavy metals: ≤2 ppm

SMILES string

BrBr

InChI

1S/Br2/c1-2

InChI key

GDTBXPJZTBHREO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bromine can be used as a brominating and oxidizing agent in chemical synthesis.
It can be used:
  • To brominate alkylbenzenes at the benzylic position using photocatalytic conditions.
  • To prepare brominated aromatic compounds by electrophilic aromatic substitution reactions in the presence of Lewis acid catalysts.
  • For the bromination of carbonyl compounds at a carbonyl group via Hell-Volhard-Zelinski reaction in the presence of phosphorus trihalides.
  • To prepare isocyanates, carbamates, or amines by reacting with primary amides in the presence of base via Hofmann rearrangement.

Other Notes

Contains organic bromine compounds

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bromine
Goehring RR
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Use of bromine and bromo-organic compounds in organic synthesis
Saikia I, et al.
Chemical Reviews, 116, 6837-7042 (2016)
John D Sivey et al.
Environmental science & technology, 47(3), 1330-1338 (2013-01-18)
HOBr, formed via oxidation of bromide by free available chlorine (FAC), is frequently assumed to be the sole species responsible for generating brominated disinfection byproducts (DBPs). Our studies reveal that BrCl, Br(2), BrOCl, and Br(2)O can also serve as brominating
Kimberly T Barrett et al.
Journal of the American Chemical Society, 135(8), 2963-2966 (2013-02-16)
We report the enantioselective synthesis of atropisomeric benzamides employing catalytic electrophilic aromatic substitution reactions involving bromination. The catalyst is a simple tetrapeptide bearing a tertiary amine that may function as a Brønsted base. A series of tri- and dibrominations were
Vitalij V Levin et al.
Organic letters, 15(4), 917-919 (2013-02-02)
Reactions of difluorocarbene with benzyl and alkylzinc halides leading to fluorinated organozinc species have been described. The generated α-difluorinated organozinc reagents are reasonably stable in solution and can be quenched with external electrophiles (iodine, bromine, proton), affording compounds containing the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service