Skip to Content
Merck
All Photos(2)

Key Documents

697079

Sigma-Aldrich

Titanium tetrachloride

packaged for use in deposition systems

Synonym(s):

Titanium(IV) chloride, TTC, Titanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl4
CAS Number:
Molecular Weight:
189.68
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

50 mmHg ( 55 °C)
9.6 mmHg ( 20 °C)

Assay

≥99.995% (trace metals analysis)

form

liquid

reaction suitability

core: titanium
reagent type: catalyst

bp

135-136 °C (lit.)

mp

−25 °C (lit.)

density

1.73 g/mL at 20 °C (lit.)

SMILES string

Cl[Ti](Cl)(Cl)Cl

InChI

1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

XJDNKRIXUMDJCW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 22 Titanium

Application

Titanium tetrachloride may be used in the vapor synthesis of titania powder by oxidaition. It may be used as a catalyst in cross and asymmetric aldol reactions. Other possible uses of titanium tetrachloride is in the synthesis of :
  • γ,δ-unsaturated alcohols from allylsilanes and carbonyl compounds,
  • N-Phosphinoylimines and N-sulphonylimines from aromatic aldehydes,
  • Homoallyl ethers from allylsilanes and acetals.6

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M T Crimmins et al.
The Journal of organic chemistry, 66(3), 894-902 (2001-06-30)
Asymmetric aldol additions using chlorotitanium enolates of N-acyloxazolidinone, oxazolidinethione, and thiazolidinethione propionates proceed with high diastereoselectivity for the Evans or non-Evans syn product depending on the nature and amount of the base used. With 1 equiv of titanium tetrachloride and
The titanium tetrachloride induced synthesis of N-phosphinoylimines and N-sulphonylimines directly from aromatic aldehydes
Jennings WB and Lovely CJ
Tetrahedron, 47(29), 5561-5568 (1991)
New cross-aldol reactions. Reactions of silyl enol ethers with carbonyl compounds activated by titanium tetrachloride
Mukaiyama T, et al.
Journal of the American Chemical Society, 96(24), 7503-7509 null
Hossein Noyan et al.
PloS one, 10(6), e0130658-e0130658 (2015-06-23)
To understand the molecular pathways underlying the cardiac preconditioning effect of short-term caloric restriction (CR). Lifelong CR has been suggested to reduce the incidence of cardiovascular disease through a variety of mechanisms. However, prolonged adherence to a CR life-style is
Ravit Madar et al.
Brain, behavior, and immunity, 48, 301-312 (2015-05-30)
Neuroinflammation in the central nervous system is detrimental for learning and memory, as evident form epidemiological studies linking developmental defects and maternal exposure to harmful pathogens. Postnatal infections can also induce neuroinflammatory responses with long-term consequences. These inflammatory responses can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service