
Fetal Bovine Serum

Synonym(s):
FBS, FCS, sera, serum
Products
Product Category
Brand
Biological Source
Product Line
Tissue
Chemical Composition
Quality Segment
Geographic Origin
Sterilization Method
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2