Skip to Content
Merck
All Photos(1)

Key Documents

F9423

Sigma-Aldrich

Fetal Bovine Serum

Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma

Synonym(s):

Fetal Calf Sera, Fetal Calf Serum, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

bovine fetus

Quality Level

sterility

sterile-filtered

product line

FBS Classic

quality

USDA approved

composition

Bovine IgG, ≤1 mg/mL
Hemoglobin, ≤20 mg/dL

origin

Australia origin

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

suitability

suitable for

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

FBS sourced from Australia is use in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture. Australia carries the best possible rating in relation toGeographical Bovine Spongiform Encephalopathy (BSE) Risk (GBR) as designated by the Scientific Steering Committee on the Geographical Risk of Bovine Spongiform Encephalopathy. The classification for Australia is GBR Level I.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Adrian Westhaus et al.
Human gene therapy, 31(9-10), 575-589 (2020-02-01)
Adeno-associated virus (AAV) vectors are quickly becoming the vectors of choice for therapeutic gene delivery. To date, hundreds of natural isolates and bioengineered variants have been reported. While factors such as high production titer and low immunoreactivity are important to
Li Dong et al.
Cell death & disease, 11(6), 442-442 (2020-06-10)
Cells from two murine lymphoid lines died 24-48 h after treatment with the glucocorticoid dexamethasone. Deletion of Bax and Bak1 prevented rapid apoptosis, but treatment with dexamethasone for greater 6 days still led to cell death that was characterized by release
Marti Cabanes-Creus et al.
Molecular therapy. Methods & clinical development, 17, 1139-1154 (2020-06-04)
Use of the prototypical adeno-associated virus type 2 (AAV2) capsid delivered unexpectedly modest efficacy in an early liver-targeted gene therapy trial for hemophilia B. This result is consistent with subsequent data generated in chimeric mouse-human livers showing that the AAV2
Suad M Abdirahman et al.
Cancers, 12(9) (2020-08-23)
Colorectal cancer (CRC) is a challenging disease, with a high mortality rate and limited effective treatment options, particularly for late-stage disease. Patient-derived xenografts (PDXs) have emerged as an informative, renewable experimental resource to model CRC architecture and biology. Here, we
Bashar M Thejer et al.
BMC molecular and cell biology, 21(1), 24-24 (2020-04-05)
Progesterone Receptor Membrane Component 1 (PGRMC1) is expressed in many cancer cells, where it is associated with detrimental patient outcomes. It contains phosphorylated tyrosines which evolutionarily preceded deuterostome gastrulation and tissue differentiation mechanisms. We demonstrate that manipulating PGRMC1 phosphorylation status

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service