Skip to Content
Merck
All Photos(1)

Key Documents

12107C

Sigma-Aldrich

Fetal Bovine Serum

USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture

Synonym(s):

Fetal Calf Sera, Fetal Calf Serum, GI FBS, GI FCS, GI Fetal Bovine Sera, GI Fetal Bovine Serum, GI Fetal Calf Serum, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

bovine fetus

sterility

sterile-filtered

product line

FBS Premier

composition

hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.

Preparation Note

Gamma irradiation by SER-TAIN process (25-40kGy).

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yusuke Ohnishi et al.
PloS one, 3(5), e2248-e2248 (2008-05-22)
Allele-specific gene silencing by RNA interference (RNAi) is therapeutically useful for specifically inhibiting the expression of disease-associated alleles without suppressing the expression of corresponding wild-type alleles. To realize such allele-specific RNAi (ASP-RNAi), the design and assessment of small interfering RNA
Feng C Zhou et al.
BMC genomics, 12, 124-124 (2011-02-23)
We have previously demonstrated that alcohol exposure at early neurulation induces growth retardation, neural tube abnormalities, and alteration of DNA methylation. To explore the global gene expression changes which may underline these developmental defects, microarray analyses were performed in a
Silvia Goldoni et al.
The American journal of pathology, 173(3), 844-855 (2008-08-09)
Decorin, a member of the small leucine-rich proteoglycan gene family, down-regulates members of the ErbB receptor tyrosine kinase family and attenuates their signaling, leading to growth inhibition. We investigated the effects of decorin on the growth of ErbB2-overexpressing mammary carcinoma
Jelle Hendrix et al.
Nucleic acids research, 39(4), 1310-1325 (2010-10-27)
Nearly all cellular and disease related functions of the transcriptional co-activator lens epithelium-derived growth factor (LEDGF/p75) involve tethering of interaction partners to chromatin via its conserved integrase binding domain (IBD), but little is known about the mechanism of in vivo
Robert C Tuckey et al.
The Journal of steroid biochemistry and molecular biology, 112(4-5), 213-219 (2008-11-13)
Cytochrome P450scc (CYP11A1) metabolizes vitamin D3 to 20-hydroxyvitamin D3 as the major product, with subsequent production of dihydroxy and trihydroxy derivatives. The aim of this study was to determine whether cytochrome P450scc could metabolize 1alpha-hydroxyvitamin D3 and whether products were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service