Skip to Content
Merck
All Photos(5)

Key Documents

TMS-016

Sigma-Aldrich

Fetal Bovine Serum

US Origin, HL-1 Cell Screened FBS, sterile-filtered, suitable for cell culture

Synonym(s):

FBS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.71
biological source:
bovine fetus
origin:
USA origin
sterility:
sterile; sterile-filtered
form:
liquid
Pricing and availability is not currently available.

biological source

bovine fetus

Quality Level

sterility

sterile; sterile-filtered

product line

FBS Select

form

liquid

origin

USA origin

technique(s)

cell culture | mammalian: suitable (cell culture)

impurities

endotoxin, tested
mycoplasma, tested
virus, tested

storage temp.

−20°C

General description

HL-1 Cell Screened Fetal Bovine Serum is exclusively of US origin. Each lot is sterile filtered and mycoplasma, endotoxin, and virus tested. Additionally, each lot is screened using HL-1 mouse cardiac muscle cells (cat. no. SCC065) to ensure maintenance of differentiated cardiomyocyte phenotype and contractile activity in vitro. A Certificate of Analysis is available upon request.
This serum has been derived from blood aseptically collected in USDA approved abattoirs in the United States of America. This serum was aseptically processed and filter sterilized in the United States. The country of origin of the the crude blood (donor cow origin) is the United States.

Application

This serum is US Origin FBS and has not been heat inactivated or gamma irradiated.

Quality

Each lot is tested and qualified for use with mouse HL-1 cardiac muscle cell line.

Storage and Stability

Frozen FBS should be stored at -20ºC until expiration date.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service