Skip to Content
Merck

Skip To

F0392

Fetal Bovine Serum

USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture

Synonym(s):

Fetal Bovine Sera, Fetal Calf Sera, FBS, FCS, sera, serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160409
NACRES:
NA.71
MDL number:
Biological source:
bovine fetus
Origin:
USA origin
Sterility:
sterile-filtered
Form:
liquid
Shipped in:
dry ice
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine fetus

Quality Segment

sterility

sterile-filtered

product line

FBS Select

form

liquid

composition

Bovine IgG, ≤1 mg/mL , Hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Application

Fetal Bovine Serum has been used as a supplement for various cell culture techniques as it provides many non-defined growth promoting and survival enhancing factor to cells in culture.

Biochem/physiol Actions

Fetal Bovine Serum (FBS) is a constituent of animal cell culture media. It contains hormones and growth factors. FBS promotes in vitro cell culture, by providing binding proteins, lipids, minerals, attachment and spreading factors.

Preparation Note

10,000 Molecular Weight Cut-off

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F676512107CF7524
form

liquid

form

-

form

-

form

-

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

biological source

bovine fetus

biological source

bovine fetus

biological source

bovine fetus

biological source

bovine fetus

origin

USA origin

origin

USA origin

origin

USA origin

origin

non-USA origin

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice


Still not finding the right product?

Explore all of our products under Fetal Bovine Serum


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Blocking anaplerotic entry of glutamine into the TCA cycle sensitizes K-Ras mutant cancer cells to cytotoxic drugs
Saqcena M, et al.
Oncogene, 34(20), 2672-2672 (2015)
Liver Tissue Engineering Based on Poly (ethylen Glycol) Hydrogels (2010)
The use of fetal bovine serum: ethical or scientific problem?
Jochems CEA, et al.
Alternatives to Laboratory Animals : ATLA, 30(2), 219-227 (2002)



Global Trade Item Number

SKUGTIN
F0392-100ML04061833611166
F0392-500ML04061838259431

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Once a bottle of Fetal Bovine Serum (FBS) is thawed, for how long can the bottle be used?

    1 answer
    1. Once Fetal Bovine Serum has been thawed, the bottle can be used in cell culture for up to 7 days for most cell types when stored  refrigerated at 2-8°C.  If the cells are not fastidous (e.g. HeLa or fibroblasts), the thawed and refrigerated serum can be used for up to 30 days.  For some extremely fastidious cell lines, it may be necessary to screen individual lots for acceptable performance.  For uses such as blocking for various immuno procedures, usage will generally be for 30 days and perhaps somewhat longer when stored at 2-8°C.  For other procedures, usage beyond 30 days storage at 2-8°C will need to be determined.

      Helpful?

Reviews

No rating value

Active Filters