Skip to Content
Merck
All Photos(3)

Key Documents

Y4645

Sigma-Aldrich

Anti-phospho-YAP1 (pTyr357) antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-YAP, Anti-YAP2, Anti-YAP65, Anti-Yes-associated protein 1, 65kDa

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~65 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using HEK-293 cells co-transfected with human YAP1 and c-Abl
western blot: 0.5-1 μg/mL using HEK-293T cells co-transfected with human YAP1 and human c-Abl

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr357)

Gene Information

chicken ... YAP1(396171)
human ... YAP1(10413)
mouse ... Yap1(22601)

General description

Yes-associated protein 1 (YAP1) is a 65 kDa and a modular adapter protein with multiple protein interaction domains that was originally identified based on its interaction with Src-family tyrosine kinase c-Yes. In addition to a SH3-binding motif, YAP1 contains a proline-rich amino terminus, a WW domain, a coiled-coil and a PDZ-binding motif at the extreme C-terminus. YAP1 binds to p73 through its WW domain and the PPPY motif of p73. This interaction is required for the ability of YAP1 to co-activate p73-responsive genes.

Specificity

Anti-phospho-YAP1 pTyr357 antibody is specific for human phospho-YAP1 pTyr357. The product may react with chicken and mouse phospho-YAP1 pTyr357. In immunoblotting, staining of the phospho-YAP1 (pTyr357) band is specifically inhibited by the immunizing peptide; but no inhibition is observed with the unmodified peptide.

Immunogen

synthetic peptide corresponding to amino acids 351-362 (pTyr357) located in the C-terminal region of human YAP1. The sequence is identical in chicken YAP1 and highly conserved in mouse YAP1 (single amino acid substitution).

Application

Anti-phospho-YAP1 (pTyr357) antibody produced in rabbit has been used in western blotting and immunofluorescence.

Biochem/physiol Actions

Inhibition of Akt potentiates the nuclear re-localization of YAP1 to induce apoptosis by p73.
YAP1 is a WW domain containing protein that is known to associate with a wide range of transcription factors and functions as a transcriptional co-activator. It may also regulate p73 gene function in response to DNA damage. Alterations in YAP1 have been associated with tumor development
Anti-phospho-YAP1 pTyr357 antibody is specific for human phospho-YAP1 pTyr357. The product may react with chicken and mouse phospho-YAP1 pTyr357. In immunoblotting, staining of the phospho-YAP1 (pTyr357) band is specifically inhibited by the immunizing peptide; but no inhibition is observed with the unmodified peptide.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conjugated Bile Acids Promote Invasive Growth of Esophageal Adenocarcinoma Cells and Cancer Stem Cell Expansion via Sphingosine 1-Phosphate Receptor 2--Mediated Yes-Associated Protein Activation
Liu R, et al.
The American Journal of Pathology, 188(9), 2042-2058 (2015)
The mammalian Hippo pathway: regulation and function of YAP1 and TAZ
Kodaka M, et al.
Cellular and Molecular Life Sciences, 72 (2015)
Akt Phosphorylates the Yes-Associated Protein, YAP, to Induce Interaction with 14-3-3 and Attenuation of p73-Mediated Apoptosis
Basu S, et al.
Molecular Cell, 11 (2003)
Akihiko Komuro et al.
The Journal of biological chemistry, 278(35), 33334-33341 (2003-06-17)
The ErbB-4 receptor protein-tyrosine kinase is proteolytically processed by membrane proteases in response to the ligand or 12-O-tetradecanoylphorbol-13-acetate stimulation resulting in the cytoplasmic fragment translocating to the cell nucleus. The WW domain-containing co-transcriptional activator Yes-associated protein (YAP) associates physically with
Fernando D Camargo et al.
Current biology : CB, 17(23), 2054-2060 (2007-11-06)
The mechanisms that regulate mammalian organ size are poorly understood. It is unclear whether the pathways that control organ size also impinge on stem/progenitor cells. A highly expressed gene in stem cells is YAP1, the ortholog of Drosophila Yorkie, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service