Skip to Content
Merck
All Photos(1)

Key Documents

Y0771

Sigma-Aldrich

Anti-phospho-YAP1 (pTyr357) antibody, Mouse monoclonal

clone PYP-76, purified from hybridoma cell culture

Synonym(s):

Anti-COB1, Anti-YAP, Anti-YAP-1, Anti-YAP2, Anti-YAP65, Anti-YKI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

PYP-76, monoclonal

form

buffered aqueous solution

mol wt

antigen ~65 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 0.5-1 μg/mL using total cell extract of HEK-293T cells co-transfected with wt YAP and a constitutively active mutant of c-Abl (Δ1-81)

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr357)

Gene Information

human ... YAP1(10413)
mouse ... Yap1(22601)

General description

Monoclonal Anti-phospho-YAP1 (pTyr357) (mouse IgM isotype) is derived from the hybridoma PYP-76 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic phosphopeptide corresponding to a fragment containing (pTyr357) of human YAP1 conjugated to KLH. Yes-associated protein 1 (YAP1) is a modular adapter protein with multiple protein interaction domains that was originally identified based on its interaction with the Src-family tyrosine kinase c-Yes. YAP1 contains a proline-rich amino terminus, a WW domain, a coiled-coil and a PDZ-binding motif at the extreme C-terminus.

Application

Anti-phospho-YAP1 (pTyr357) antibody, Mouse monoclonal has been used in
  • immunoblotting
  • enzyme linked immunosorbent assay (ELISA)
  • immunofluorescence

Biochem/physiol Actions

YAP1 has been shown to stabilize p73 by preventing Itch-mediated ubiquitination of p73.
YAP1 is a WW domain containing protein that is known to associate with a wide range of transcription factors and functions as a transcriptional co-activator. It may also regulate p73 gene function in response to DNA damage. Alterations in YAP1 have been associated with tumor development

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conjugated Bile Acids Promote Invasive Growth of Esophageal Adenocarcinoma Cells and Cancer Stem Cell Expansion via Sphingosine 1-Phosphate Receptor 2-Mediated Yes-Associated Protein Activation
Liu R, et al.
The American Journal of Pathology, 188(9), 2042-2058 (2018)
Xiuye Jin et al.
Cell death & disease, 15(2), 148-148 (2024-02-16)
Metastasis accounts for the majority of cases of cancer recurrence and death in patients with advanced non-small cell lung cancer (NSCLC). Farnesoid X Receptor (FXR) is a bile acid nuclear receptor that was recently found to be upregulated in NSCLC
Ting-Fang Lee et al.
Oncotarget, 8(52), 89539-89551 (2017-11-23)
Epidermal growth factor receptor (EGFR) mutations are found in lung adenocarcinomas leading to tumor cells proliferation and survival. EGFR tyrosine kinase inhibitors (TKIs) that block EGFR activity are effective therapeutics for EGFR-mutant lung adenocarcinoma patients, but TKI-resistance inevitably occurs. The
The Yes-associated protein 1 stabilizes p73 by preventing Itch-mediated ubiquitination of p73
Levy D, et al.
Cell Death and Differentiation, 14(4), 743-743 (2007)
Akihiko Komuro et al.
The Journal of biological chemistry, 278(35), 33334-33341 (2003-06-17)
The ErbB-4 receptor protein-tyrosine kinase is proteolytically processed by membrane proteases in response to the ligand or 12-O-tetradecanoylphorbol-13-acetate stimulation resulting in the cytoplasmic fragment translocating to the cell nucleus. The WW domain-containing co-transcriptional activator Yes-associated protein (YAP) associates physically with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service