Skip to Content
Merck
All Photos(1)

Key Documents

T8885

Sigma-Aldrich

Thrombin from human plasma

lyophilized powder, Suitable for routine use in the thrombin time test

Synonym(s):

Factor IIa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

human plasma

form

lyophilized powder

packaging

vial of ≥10 NIH units

impurities

HIV, hepatitis B and hepatitis C, tested negative

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... F2(2147)

Looking for similar products? Visit Product Comparison Guide

General description

Thrombin is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects.

Application

Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags. Thrombin has been used in a study that compared efficacy and safety of low-molecular-weight heparins (LMWHs) to other anticoagulants as venous thromboembolism prophylaxis in major orthopedic surgery.

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Unit Definition

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard

Reconstitution

When reconstituted with 1 ml H2O, one vial will contain 10 mg bovine serum albumin, 0.15 M NaCl and 0.05 M sodium citrate, pH 6.5

Analysis Note

The NIH assay procedure uses 0.2 ml diluted plasma (1:1 with saline) as a substrate and 0.1ml of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana M Sobieraj et al.
Pharmacotherapy, 32(9), 799-808 (2012-06-30)
To evaluate the comparative efficacy and safety of low-molecular-weight heparins (LMWHs) versus other anticoagulants as venous thromboembolism prophylaxis in major orthopedic surgery. Systematic review with meta-analysis of 37 randomized controlled trials. Patients undergoing total hip replacement, total knee replacement, or
Meihua Hao et al.
Reproductive biology and endocrinology : RB&E, 20(1), 85-85 (2022-06-07)
Women with endometriosis have been shown to have a reduced vagal tone as compared with controls and vagotomy promoted while vagus nerve stimulation (VNS) decelerated the progression of endometriosis in mice. Extensive research also has shown that the activation of
Shenghui Huang et al.
Reproductive sciences (Thousand Oaks, Calif.), 29(4), 1170-1187 (2022-02-01)
The development of more efficacious, non-hormonal therapeutics for endometriosis is still an unmet medical need begging to be fulfilled. Growing evidence indicates that endometriotic lesions are wounds undergoing repeated tissue injury and repair, and, as such, platelets play an important
Nathalie Busschaert et al.
Chembiochem : a European journal of chemical biology, 23(6), e202100670-e202100670 (2022-01-06)
The thrombin binding aptamer (TBA) is a 15-mer DNA oligonucleotide (5'-GGT TGG TGT GGT TGG-3'), that can form a stable intramolecular antiparallel chair-like G-quadruplex structure. This aptamer shows anticoagulant properties by interacting with one of the two anion binding sites
Antonella Virgilio et al.
Chembiochem : a European journal of chemical biology, 15(16), 2427-2434 (2014-09-13)
We report an investigation into analogues of the thrombin binding aptamer (TBA). Individual thymidines were replaced by the unusual residue 5-hydroxymethyl-2'-deoxyuridine (hmU). This differs from the canonical thymidine by a hydroxyl group on the 5-methyl group. NMR and CD data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service