Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thrombin from human plasma
Synonym(s):
Factor IIa
CAS Number:
9002-04-4
EC Number:
232-648-7
Products
Brand
Sigma-Aldrich (6)
Biological Source
Gene Alias
Physical Form
Tissue
Quality Segment
Application
Package Size
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
T6884
Thrombin from human plasma,
lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)
Expand
View Pricing
T7009
Thrombin from human plasma,
lyophilized powder, ≥1,000 NIH units/mg protein (E1%/280, 18.3)
Expand
View Pricing
T8885
Thrombin from human plasma,
lyophilized powder, Suitable for routine use in the thrombin time test
Expand
View Pricing
T1063
Thrombin from human plasma,
lyophilized powder, ≥2800 NIH units/mg protein (E1%/280, 18.3)
Expand
View Pricing
T4393
Thrombin from human plasma,
lyophilized powder, 1500-3500 NIH units/mg protein (E1%/280, 18.3), suitable for cell culture
Expand
View Pricing
T7572
Thrombin from human plasma,
400-1000 NIH units/mg protein
Expand
View Pricing