Skip to Content
Merck
All Photos(3)

Key Documents

T0440

Sigma-Aldrich

3,3′,5,5′-Tetramethylbenzidine

peroxidase substrate, chromogenic, liquid

Synonym(s):

TMB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.83

Product Name

3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate

Quality Level

form

liquid

storage temp.

2-8°C

General description

TMB substrate is a chromogenic substrate for horseradish peroxidase - HRP.

Application

TMB substrate is used in ELISA procedures, which utilize horseradish peroxidase conjugates. TMB substrate develops a soluble blue reaction product that can be read at 370 or 655 nm at 370 or 655 nm. The reaction is stopped with acid, forming a yellow reaction product which enables accurate intensity measurement at 450 nm.

How to use our TMB substrate:
  • Our TMB substrate is supplied as a ready-to-use peroxidase substrate containing TMB in a mildly acidic buffer. Prior to reaction with peroxidase, the substrate should be a colorless to light bluish-green solution. The substrate system develops a blue reaction product when reacted with peroxidase.
  • Upon reaching your assay endpoint with addition of acid, measure the TMB substrate intensity at 450 nm.
  • Our TMB Substrate is recommended for ELISA (microwell) procedures, and has been used in various assays, as a substrate for horseradish peroxidase antibody conjugate for detecting hendra virus glycoprotein, tetracosactide, sIgA and neutrophil extracellular traps (NETs).
  • 3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System is not recommended for membrane applications. Since TMB substrate produces a soluble reaction product, it is not recommended for histochemistry or blotting.
Note:TMB substrate is light-sensitive and should be protected from direct sunlight or UV sources.

recommended

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise Traxler et al.
Clinica chimica acta; international journal of clinical chemistry, 473, 127-132 (2017-08-29)
Heat shock proteins (HSPs) represent intracellular mechanisms of stress response. Clinical implications of their (systemic) expression in patients with chronic heart failure (HF) remain inconclusive. In outpatients with chronic stable HF plasma HSP27 levels were measured using ELISA. Patients were
Myeloid-specific deletion of peptidylarginine deiminase 4 (PAD4) mitigates atherosclerosis.
Liu Y, et al.
Frontiers in immunology, 9(4), 1680-1680 (2018)
Spontaneous and tetracosactide-induced anti-ACTH antibodies in man
Gan EH, et al.
Clinical Endocrinology, 84(4), 489-495 (2016)
R Roth et al.
Journal of applied microbiology, 122(5), 1402-1411 (2017-03-10)
To investigate the production of soluble cross-reacting material 197 (CRM The use of co-expression of a sulphydryl oxidase (SOX) and protein disulphide isomerase for the production of soluble CRM SOX and protein disulphide isomerase are enzymes involved in the formation
A network approach for provisional assay recognition of a Hendra virus antibody ELISA: test validation with low sample numbers from infected horses
Colling A, et al.
Journal of Veterinary Diagnostic Investigation : Official Publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 30(3), 362-369 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service