Skip to Content
Merck
All Photos(1)

Key Documents

T7572

Sigma-Aldrich

Thrombin from human plasma

400-1000 NIH units/mg protein

Synonym(s):

Factor IIa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

specific activity

400-1000 NIH units/mg protein

impurities

HIV, hepatitis B and hepatitis C, tested negative

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... F2(2147)

Looking for similar products? Visit Product Comparison Guide

General description

Lyophilized powder containing sucrose, sodium chloride and Tris.
Thrombin is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects. [1]

Application

Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags. Thrombin has been used in a study to assess an innovative flow cytometric approach for small-size platelet microparticles. [2]

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Unit Definition

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard

Analysis Note

The NIH assay procedure uses 0.2 ml diluted plasma (1:1 with saline) as a substrate and 0.1ml of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

At the cutting edge: conformational PARtiality.
T Madhusudhan et al.
Journal of thrombosis and haemostasis : JTH, 10(8), 1672-1674 (2012-06-30)
Silvia Montoro-García et al.
Thrombosis and haemostasis, 108(2), 373-383 (2012-06-29)
Microparticles (MPs) are small submicron membrane-derived vesicles shed from a variety of cells and they have been implicated in different disorders. Accordingly, understanding of physiological characteristics of MPs and improvement of methods of their quantification are important for further advance
Walter-Michael Halbmayer et al.
Clinical chemistry and laboratory medicine, 50(9), 1601-1605 (2012-10-27)
Dabigatran etexilate is a new oral anticoagulant for the therapy and prophylaxis of venous thromboembolism and stroke prevention in patients with atrial fibrillation. To investigate the extent of interactions of this new anticoagulant with frequently used coagulation assays, we completed
Herbert Nar
Trends in pharmacological sciences, 33(5), 279-288 (2012-04-17)
The quest for novel medications to treat thromboembolic disorders such as venous thrombosis, pulmonary embolism and stroke received a boost when the 3D structures of two major players in the blood coagulation cascade were determined in 1989 and 1993. Structure-guided
Lukas Graf et al.
Swiss medical weekly, 142, w13684-w13684 (2012-10-06)
New oral anticoagulants used as single target inhibitors of coagulation enzymes have been developed and tested in extensive trial programmes. Results of most of these trials showed non-inferior and/or superior efficacy and safety compared to standard treatment with LMWH or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service