T8665
3,3′,5,5′-Tetramethylbenzidine
peroxidase substrate, chromogenic, liquid
Synonym(s):
TMB
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.83
Recommended Products
Product Name
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System, peroxidase substrate
Quality Level
form
liquid
storage condition
protect from light
shipped in
wet ice
storage temp.
2-8°C
Related Categories
Application
TMB is a substrate for horseradish peroxidase. Forms a soluble blue end product that may be read at 370 nm or 655 nm. The TMB reaction may be stopped with 0.5 M H2SO4, resulting in a yellow color, and read at 450 nm. Recommended for ELISA (microwell) procedures, not recommended for membrane applications.
3,3′,5,5′-Tetramethylbenzidine has been used in a study to demonstrate that by measuring substrate absorption values off the absorption maximum, the measuring range of any immunoenzymometric assay can be extended significantly without jeopardizing assay characteristics. 3,3′,5,5′-Tetramethylbenzidine has also been used in a study to understand the formation of CuS concave superstructures with peroxidase-like activity.
3,3′,5,5′-Tetramethylbenzidine has been used in a study to demonstrate that by measuring substrate absorption values off the absorption maximum, the measuring range of any immunoenzymometric assay can be extended significantly without jeopardizing assay characteristics. 3,3′,5,5′-Tetramethylbenzidine has also been used in a study to understand the formation of CuS concave superstructures with peroxidase-like activity.
Caution
This substrate is very light-sensitive and should be protected from direct sunlight or UV sources.
Physical form
Ready-to-use.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 1
Target Organs
Eyes,Central nervous system
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Paweł Kozielewicz et al.
Molecular pharmacology, 97(1), 23-34 (2019-11-11)
Smoothened (SMO) is a GPCR that mediates hedgehog signaling. Hedgehog binds the transmembrane protein Patched, which in turn regulates SMO activation. Overactive SMO signaling is oncogenic and is therefore a clinically established drug target. Here we establish a nanoluciferase bioluminescence
Sui-Yuan Chang et al.
Scientific reports, 11(1), 8692-8692 (2021-04-24)
A metal nanoparticle composite, namely TPNT1, which contains Au-NP (1 ppm), Ag-NP (5 ppm), ZnO-NP (60 ppm) and ClO2 (42.5 ppm) in aqueous solution was prepared and characterized by spectroscopy, transmission electron microscopy, dynamic light scattering analysis and potentiometric titration. Based on the in
S Madersbacher et al.
Methods (San Diego, Calif.), 21(1), 41-50 (2000-04-15)
As a glycoprotein hormone, human chorionic gonadotropin (hCG) is not a single molecular entity. This term comprises not only the bioactive heterodimer hCG but also an array of molecular protein backbone and glycosylation variants, such as its free beta (hCGbeta)
Belén Morón et al.
PloS one, 3(5), e2294-e2294 (2008-05-30)
Celiac disease is a permanent intolerance to gluten prolamins from wheat, barley, rye and, in some patients, oats. Partially digested gluten peptides produced in the digestive tract cause inflammation of the small intestine. High throughput, immune-based assays using monoclonal antibodies
Tomasz Brudek et al.
Molecular neurodegeneration, 12(1), 44-44 (2017-06-09)
Parkinson's' disease (PD) and Multiple System Atrophy (MSA) are progressive brain disorders characterized by intracellular accumulations of α-synuclein and nerve cell loss in specific brain areas. This loss causes problems with movement, balance and/or autonomic functions. Naturally occurring autoantibodies (NAbs)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service