T0565
3,3′,5,5′-Tetramethylbenzidine
TMB membrane substrate, chromogenic, liquid
Synonym(s):
TMB membrane substrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.83
Recommended Products
Product Name
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for Membranes, ready to use solution
Quality Level
form
liquid
storage temp.
2-8°C
Related Categories
General description
3,3′,5,5′-Tetramethylbenzidine (TMB) is a chromogenic substrate for horseradish peroxidase (HRP) conjugates. It develops a permanent, insoluble, dark blue reaction product and is useful in colorimetric quantification.
Application
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for Membranes has been used as a chromogenic substrate in western blot analysis. It has also been used as a liquid substrate in enzyme-linked immunosorbent assay (ELISA).
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for Membranes has been used in:
- enzyme-linked immune absorbent spot (ELISPOT) assay
- as a substrate for IgG peroxidase
- in the visualization of immunocomplexes
Physical form
Ready-to-use.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Effects of nerve growth factor antagonist K252a on peritoneal mast cell degranulation: implications for rat postoperative ileus
Berdun S, et al.
American Journal of Physiology: Gastrointestinal and Liver Physiology, 309(10), G801-G806 (2015)
Navilla Apú et al.
Open life sciences, 18(1), 20220577-20220577 (2023-08-17)
Most laboratory tests to detect the presence of anti-SARS-CoV-2 antibodies use enzyme-linked immunosorbent assays (ELISA) or chemiluminescence immunoassays (CLIA); however, equipment for these immunoassays is unavailable in many areas of low- and middle-income countries. Rapid lateral flow immunoassay (LFIA) tests
Hesham Saeed et al.
Preparative biochemistry & biotechnology, 52(6), 668-680 (2021-10-07)
Microbial L-asparaginases are aminohydrolases that hydrolyze L-asparagine to L-aspartate. They are used to treat acute lymphoblastic leukemia and Hodgkin's lymphomas and in food industries. Increasing demand for L-ASNases is therefore needed. In the current study, the recombinant L-ASNase from Dickeya
Testin (TES) as a candidate tumour suppressor and prognostic marker in human astrocytoma
Steponaitis G, et al.
Oncology Letters, 12(5), 3305-3311 (2016)
Srdjan M Dragovic et al.
Cell host & microbe, 23(4), 523-535 (2018-04-13)
Plasmodium infection begins with the bite of an anopheline mosquito, when sporozoites along with saliva are injected into a vertebrate host. The role of the host responses to mosquito saliva components in malaria remains unclear. We observed that antisera against
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service