T5569
3,3′,5,5′-Tetramethylbenzidine
peroxidase substrate, chromogenic, liquid
Synonym(s):
TMB substrate for ELISA (Super Slow)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
3,3′,5,5′-Tetramethylbenzidine Liquid Substrate, Super Slow, for ELISA, peroxidase substrate
form
liquid
shipped in
wet ice
storage temp.
2-8°C
General description
3,3′,5,5′-Tetramethylbenzidine (TMB) is an electrochemical substrate for low level detection of horse radish peroxidase (HRP). It is a highly sensitive and dual function substrate, since it produces both soluble and insoluble reaction products. It has rapid reaction rate and it is suitable for on-line kinetic analysis. It does not possess carcinogenic properties.
Application
3,3′,5,5′-Tetramethylbenzidine Liquid Substrate, Super Slow, for ELISA has been used as a substrate for enzyme linked immunosorbent assay (ELISA).
Contains TMB in a mildly acidic buffer. The substrate is supplied as a one component ready to use solution. Rate kinetics are approx. 60% slower than traditional TMB formulations. Not recommended for membrane or immunohistochemical applications that require a precipitating reaction product.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
PloS one, 17(10), e0276107-e0276107 (2022-10-19)
The blood-brain barrier (BBB) presents a major obstacle in developing specific diagnostic imaging agents for many neurological disorders. In this study we aimed to generate single domain anti-mouse transferrin receptor antibodies (anti-mTfR VHHs) to mediate BBB transcytosis as components of
Soluble amyloid-beta buffering by plaques in Alzheimer disease dementia versus high-pathology controls
PLoS ONE, 13(7), e0200251-e0200251 (2018)
Pan-CC chemokine neutralization restricts splenocyte egress and reduces inflammation in a model of arthritis
Journal of Immunology, 185(4), 2544-2554 (2010)
Enzyme-linked Immunosorbent Assay (ELISA): From A to Z, 39-40 (2017)
Antibodies (Basel, Switzerland), 9(2) (2020-05-15)
Human cathelicidin LL37 is a cationic antimicrobial peptide active against bacteria and viruses and exerting immune modulatory functions. LL37 can be also a target of autoreactive B- and T-lymphocytes in autoimmune settings. Irreversible post-translational modifications, such as citrullination and carbamylation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service