Skip to Content
Merck
All Photos(4)

Documents

T0772

Sigma-Aldrich

TES sodium salt

≥99% (titration)

Synonym(s):

Sodium 2-((1,3-dihydroxy-2-(hydroxymethyl)-propan-2-yl)amino)ethane sulfonate, TES NA, N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14NNaO6S
CAS Number:
Molecular Weight:
251.23
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99% (titration)

form

crystalline powder

impurities

≤7% water content (Karl Fischer)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

water: 0.333 g/mL, clear to slightly hazy, colorless

cation traces

Pb: ≤5 ppm

application(s)

diagnostic assay manufacturing

SMILES string

[Na+].OCC(CO)(CO)NCCS([O-])(=O)=O

InChI

1S/C6H15NO6S.Na/c8-3-6(4-9,5-10)7-1-2-14(11,12)13;/h7-10H,1-5H2,(H,11,12,13);/q;+1/p-1

InChI key

GUXMFAHOYNRHBI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

TES sodium salt belongs to the category of zwitterionic buffers that contain ionizable groups of both positive and negative charges. It has a high water solubility and can effectively stabilize pH levels within the physiological pH range of 6.8 to 8.2 over a broad range of temperatures. TES sodium salt is used in the preparation of electrophoresis buffers. It is one of the commonly used Good buffers in biochemistry and molecular biology research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TES buffer-induced phase separation of aqueous solutions of several water-miscible organic solvents at 298.15 K: phase diagrams and molecular dynamic simulations
Taha M and Jer Lee M
Journal of Chemical Physics, 138(24), 244501-244501 (2013)
The use of gel and capillary electrophoresis to investigate some of the fundamental physical properties of DNA
Stellwagen N, et al.
Electrophoresis, 23(2), 167-175 (2002)
R Nakon et al.
Science (New York, N.Y.), 221(4612), 749-750 (1983-08-19)
Metal-ion affinity (formation) constants were determined for two "Good's" buffers, N-tris(hydroxymethyl)methyl-2-aminoethanesulfonic acid (TES) and N,N-bis(2-hydroxyethyl)glycine (bicine). The metal chelates formed undergo loss of an internal ligand (alcohol) proton (bicine) and undergo hydrolysis (bicine and TES) and dimerization reactions (TES). Bicine
A T Palasz et al.
Theriogenology, 70(9), 1461-1470 (2008-08-05)
The effect of the zwitterionic buffers HEPES, TES and MOPS and of PBS used for out-of-incubator procedures during standard in vitro embryo production on bovine oocytes and embryo development, morphology and on the expression patterns of eight selected genes: Fgf-4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service