Skip to Content
Merck
All Photos(1)

Documents

T6424

Sigma-Aldrich

Trypsin from human pancreas

salt-free, lyophilized powder, vial of ≥1,000 BAEE units

Synonym(s):

Serine Protease 1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

human pancreas

Quality Level

form

salt-free, lyophilized powder

packaging

vial of ≥1,000 BAEE units

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

The trypsin molecule has two domains: one is related to the enzyme active site and the tryptophan residues; the other is related to the 8-anilinonaphthalene-1-sulfonate binding.
Trypsin is a serine protease. The pancreas produce the precursor trypsinogen and secretes it to the intestine. Trypsinogen is converted to trypsin by enterokinase in the intestinal mucous membrane.

Application

Trypsin has been used in a study to assess the similarities between the hepatitis E virus and human astrovirus. Trypsin has also been used in a study to characterize a unique technique for culturing primary adult human epithelial progenitor, or stem, cells.

Unit Definition

One BAEE unit will produce a ΔA253 of 0.001 per min at pH 7.6 at 25 °C using BAEE as substrate. Reaction volume = 3.2 mL (1 cm light path).

Quantity

Minimum 1,000 BAEE units/vial

Analysis Note

Protein determined by E1%/280.

Other Notes

View more information on trypsin at www.sigma-aldrich.com/enzymeexplorer

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Liu et al.
Nature protocols, 16(7), 3522-3546 (2021-06-06)
Cellular heterogeneity is pervasive and of paramount importance in biology. Single-cell analysis techniques are indispensable for understanding the heterogeneity and functions of cells. Low-copy-number proteins (fewer than 1,000 molecules per cell) perform multiple crucial functions such as gene expression, cellular
Kelly A Dryden et al.
Journal of molecular biology, 422(5), 650-658 (2012-06-30)
Human astroviruses (HAstVs) are a major cause of gastroenteritis. HAstV assembles from the structural protein VP90 and undergoes a cascade of proteolytic cleavages. Cleavage to VP70 is required for release of immature particles from cells, and subsequent cleavage by trypsin
Industrial Enzymes and Their Applications (1998)
Cynthia Luz Marcelo et al.
BMC dermatology, 12, 8-8 (2012-06-26)
Primary keratinocytes derived from epidermis, oral mucosa, and urothelium are used in construction of cell based wound healing devices and in regenerative medicine. This study presents in vitro technology that rapidly expands keratinocytes in culture by growing monolayers under large
R J Read et al.
Biochemistry, 23(26), 6570-6575 (1984-12-18)
The X-ray crystal structure of Streptomyces griseus trypsin has been solved and refined at 1.7-A resolution. The structure of this protein had been predicted in two models on the basis of its expected homology to structures of bovine trypsin and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service