Skip to Content
Merck
All Photos(2)

Documents

B4500

Sigma-Aldrich

Nα-Benzoyl-L-arginine ethyl ester hydrochloride

trypsin substrate, chromogenic, ≥97% (HPLC), powder

Synonym(s):

BAEE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N4O3 · HCl
CAS Number:
Molecular Weight:
342.82
Beilstein:
3781694
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

Nα-Benzoyl-L-arginine ethyl ester hydrochloride, trypsin substrate

Assay

≥97% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

Cl[H].CCOC(=O)[C@H](CCCNC(N)=N)NC(=O)c1ccccc1

InChI

1S/C15H22N4O3.ClH/c1-2-22-14(21)12(9-6-10-18-15(16)17)19-13(20)11-7-4-3-5-8-11;/h3-5,7-8,12H,2,6,9-10H2,1H3,(H,19,20)(H4,16,17,18);1H/t12-;/m0./s1

InChI key

HIXDELXKSSLIKB-YDALLXLXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nα-Benzoyl-L-arginine ethyl ester hydrochloride is a substrate for trypsin. It is a nonprotein substrate used in spectroscopic assays.

Application

  • N

  • α-Benzoyl-L-arginine ethyl ester hydrochloride has been used as a substrate:
  • in peptidyl arginine deiminase (PPAD) assay in P. gingivalis vesicles
  • for assaying proteolytic activity of enzymatic extracts of papaya plant parts
  • in trypsin activity assay of the tail tendon fascicles from rat

Substrates

The prototype substrate for trypsin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optima of Trypsin-Catalyzed Hydrolysis and Its Inhibition Determined by SDS-PAGE
Zhou X, et al.
Advances in Enzyme Research, 4(01), 1-1 (2016)
Chensong Pan et al.
Analytical and bioanalytical chemistry, 387(1), 193-204 (2006-11-07)
Matrix-assisted laser desorption/ionization-time-of-flight mass spectrometry (MALDI-TOF-MS) is widely used in a variety of fields because it has the characteristics of speed, ease of use, high sensitivity, and wide detectable mass range for obtaining molecular weights and for structural characterization of
Ying-Qing Yu et al.
Analytical chemistry, 75(21), 6023-6028 (2003-11-01)
Improved in-solution tryptic digestion of proteins in terms of speed and peptide coverage was achieved with the aid of a novel acid-labile anionic surfactant (ALS). Unlike SDS, ALS solubilizes proteins without inhibiting trypsin or other common endopeptidases activity. Trypsin activity
Mohammad K Hossain et al.
Colloids and surfaces. B, Biointerfaces, 181, 85-93 (2019-05-28)
Electric current responsive magnetic composite particles are prepared in three steps. In the first step, spherical and mesoporous submicrometer-sized magnetic iron oxide (Fe3O4) core particles are prepared by solvothermal method. Then magnetic Fe3O4 particles are functionalized with amine groups using
Kinetic and Structural Differentiation of Trypsin from Different Origins
Aithal M, et al.
BioPharm International, 30(1), 38-+-38-+ (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service