Skip to Content
Merck

Skip To

T2011

Trypsin inhibitor

powder

Synonym(s):

Ovomucoid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77
EC Number:
232-906-9
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Trypsin inhibitor from chicken egg white, Type III-O (free of ovoinhibitor)

biological source

chicken egg white

Quality Segment

type

Type III-O (free of ovoinhibitor)

form

powder

mol wt

27 kDa

solubility

water: soluble 10 g/L

storage temp.

2-8°C

General description

Ovomucoid or trypsin inhibitor is an abundant protein in most avian egg whites. The hen′s egg protein is composed of about 186 amino acid residues, and is highly glycosylated. Three tandem domains are each homologous with pancreatic trypsin inhibitor. It is highly immunogenic, and probably accounts for most cases of egg allergy.

Application

Trypsin inhibitor from chicken egg white has been used:
  • to terminate proteolysis reaction
  • in OVO-1 solution to triturate the cell suspension for the dissociation of retina
  • in phosphate buffered saline for dissociation of cortices

Biochem/physiol Actions

Ovomucoid possesses trypsin inhibitor activity. It has several conformational and linear epitopes that can be bound by immunoglobulin E (IgE). It is highly stable against heat and proteolysis.

Other Notes

One trypsin unit will produce a ΔA253 of 0.001 per min with BAEE as substrate at pH 7.6 at 25 °C; reaction volume 3.2 mL, 1 cm light path.
View more information on Trypsin Inhibitor.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T9253650357T2327
biological source

chicken egg white

biological source

chicken egg white

biological source

-

biological source

Glycine max (soybean)

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

200

form

powder

form

powder

form

lyophilized solid

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

water: soluble 10 g/L

solubility

water: soluble 10 g/L

solubility

67 mM potassium phosphate buffer, pH 7.6: 5 mg/mL

solubility

-

mol wt

27 kDa

mol wt

28 kDa

mol wt

-

mol wt

-


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the difference between the Type III-O free of Ovoinhibitor in product T2011. What is availability of the material?

    1 answer
    1. The T2011 product does not contain ovoinhibitor, while the T9253 product contains ovoinhibitor, which results in chymotrypsin inhibition. For information regarding the availability of this product, please contact local customer service.

      Helpful?

Reviews

No rating value

Active Filters