Skip to Content
Merck
All Photos(1)

Key Documents

D5905

Sigma-Aldrich

3,3′-Diaminobenzidine tetrahydrochloride

peroxidase substrate, chromogenic, tablet

Synonym(s):

3,3′,4,4′-Biphenyltetramine tetrahydrochloride, 3,3′,4,4′-Tetraaminobiphenyl tetrahydrochloride, DAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2)2C6H3C6H3(NH2)2 · 4HCl
Molecular Weight:
360.11
Beilstein:
1212988
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet

form

tablet

solubility

water: 1 tablet/15 mL (may contain small insoluble particles)

storage temp.

−20°C

SMILES string

Cl.Cl.Cl.Cl.Nc1ccc(cc1N)-c2ccc(N)c(N)c2

InChI

1S/C12H14N4.4ClH/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8;;;;/h1-6H,13-16H2;4*1H

InChI key

KJDSORYAHBAGPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Specificity

Peroxidase substrate suitable for use in immunoblotting and immunohistological staining procedures. This substrate produces an insoluble end product that is brown in color and can be observed visually.

Application

3,3′-Diaminobenzidine tetrahydrochloride has been used for immunohistochemistry.
Reagent for spectrophotometric determination of selenium.

Biochem/physiol Actions

3,3′-Diaminobenzidine tetrahydrochloride is a chromogenic substrate, which is commonly used in immunoperoxidase tests.

Reconstitution

Dissolve 1 tablet in 15 ml of Tris-buffered saline, pH 7.6. Add 12 μl of fresh 30% hydrogen peroxide. DAB and hydrogen peroxide concentration may be adjusted to suit individual applications.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deletion of Dtnbp1 in mice impairs threat memory consolidation and is associated with enhanced inhibitory drive in the amygdala
Huang CCY, et al.
Translational Psychiatry, 9(1), 132-132 (2019)
Neuroprotection Induced by Energy and Protein-Energy Undernutrition Is Phase-Dependent After Focal Cerebral Ischemia in Mice
de Carvalho TS, et al.
Translational Stroke Research, 1-12 (2019)
Leonardo Della Salda et al.
Veterinary and comparative oncology, 17(3), 298-307 (2019-02-16)
Nectin-4 is an E-cadherin-based adherens junction protein of normal epithelial cells, as well as a potent mediator of anchorage-independent cancer colony formation. It is considered a tumour-associated histological and serological marker in various human cancers. The transcription factor p63 is
Alfons Torrego et al.
Thorax, 62(4), 307-313 (2007-01-26)
Airway wall remodelling and inflammation are features of chronic asthma. Transforming growth factor beta (TGF-beta) has been implicated in these processes. To determine the effect of allergen challenge on airway inflammation and remodelling and whether TGF-beta isoforms and the Smad
Enoch Muyanja et al.
The Journal of clinical investigation, 124(7), 3147-3158 (2014-06-10)
Defining the parameters that modulate vaccine responses in African populations will be imperative to design effective vaccines for protection against HIV, malaria, tuberculosis, and dengue virus infections. This study aimed to evaluate the contribution of the patient-specific immune microenvironment to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service