Skip to Content
Merck
All Photos(4)

Key Documents

P3556

Sigma-Aldrich

L-α-Phosphatidylcholine

egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
5209585
EC Number:
UNSPSC Code:
51321705
NACRES:
NA.25

biological source

egg yolk

Quality Level

type

Type XVI-E

Assay

≥99% (TLC)

form

lyophilized powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphatidylcholine belongs to the class of glycerophoshpolipids and contains choline as the head-group. Choline is attached to the glycerol of fatty acids ester-bound to backbone. It is the major phospholipid found in eukaryotic organism.

Application

L-α-Phosphatidylcholine has been used:
  • to form a thin lipid film used in Kupffer cell depletion by liposome-encapsulated clodronate
  • as a component of assay buffer used in γ-secretase in vitro assay
  • in the preparation of substrate mix for its use in HPLC (high performance liquid chromatography) assay of 5-lipoxygenase enzyme activity

Biochem/physiol Actions

A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.
It also acts as a source of lipid messengers/ bioactive lipids including: lysophosphatidylcholine, diacylglycerol, phosphatidic acid, lysophosphatidylcholine, arachidonic acid and platelet activating factor. Phosphatidylcholine is produced in the liver by the CDP-choline (cytidine diphosphocholine) pathway.

Packaging

Packaged under Argon

Preparation Note

Prepared by a modification of the procedure of Singleton, W.S., et al., J. Am. Oil Chem. Soc., 42, 53 (1965).

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marine omega-3 phospholipids: metabolism and biological activities
Burri L, et al.
International Journal of Molecular Sciences, 13(11), 15401-15419 (2012)
Ireos Filipuzzi et al.
PLoS genetics, 12(11), e1006374-e1006374 (2016-11-18)
Invasive infections by fungal pathogens cause more deaths than malaria worldwide. We found the ergoline compound NGx04 in an antifungal screen, with selectivity over mammalian cells. High-resolution chemogenomics identified the lipid transfer protein Sec14p as the target of NGx04 and
Chemical cross-linking provides a model of the gamma-secretase complex subunit architecture and evidence for close proximity of the C-terminal fragment of presenilin with APH-1
Steiner H, et al.
The Journal of Biological Chemistry, 283(50), 34677-34686 (2008)
Li Zhou et al.
Journal of agricultural and food chemistry, 65(6), 1229-1238 (2017-01-24)
Purification, characterization, and antioxidative activity in vitro of shrimp phosphatidylcholines (PCs) were investigated. The molecular structures of shrimp PCs were determined by MALDI-TOF/TOF MS. The MS
Coactosin-like protein supports 5-lipoxygenase enzyme activity and up-regulates leukotriene A4 production
Rakonjac M, et al.
Proceedings of the National Academy of Sciences of the USA, 103(35), 13150-13155 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service