Skip to Content
Merck
All Photos(1)

Key Documents

SML1558

Sigma-Aldrich

Yoda1

≥98% (HPLC), powder, Piezo1 agonist

Synonym(s):

2-[5-[[(2,6-Dichlorophenyl)methyl]thio]-1,3,4-thiadiazol-2-yl]-pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8Cl2N4S2
CAS Number:
Molecular Weight:
355.27
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Yoda1, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(nnc1c3nccnc3)SCc2c(cccc2Cl)Cl

InChI

1S/C13H8Cl2N4S2/c14-9-2-1-3-10(15)8(9)7-20-13-19-18-12(21-13)11-6-16-4-5-17-11/h1-6H,7H2

InChI key

BQNXBSYSQXSXPT-UHFFFAOYSA-N

General description

Yoda1 is a novel synthetic small molecule developed to study the kinetics, regulation and function of the cation channel Piezo-type mechanosensitive ion channel component 1 (Piezo1).

Application

Yoda1 has been used to investigate its effect on heterotrimeric Piezo1 channels.

Biochem/physiol Actions

Yoda1 is a specific activator of mouse and human cation channel Piezo1, a mechanosensitive channel that mediates Ca2+ influx on mechanical activation and has been linked to a role in blood vessel development. Yoda1 stabilizes the Piezo1 open channel. In red blood cells, this was followed by downstream activation of the KCa3.1 Gardos channel with resultant calcium influx and potassium and water efflux, causing shrinkage of the cells. Yoda1 is the first Piezo1 agonist, giving the first evidence of non-mechanical activation of a Piezo channel.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerome J Lacroix et al.
Nature communications, 9(1), 2029-2029 (2018-05-26)
Piezo proteins are transmembrane ion channels which transduce many forms of mechanical stimuli into electrochemical signals. Their pore, formed by the assembly of three identical subunits, opens by an unknown mechanism. Here, to probe this mechanism, we investigate the interaction
Chemical gating of the mechanosensitive Piezo1 channel by asymmetric binding of its agonist Yoda1.
Lacroix JJ, et al.
bioRxiv, 169516-169516 (2017)
Erika Sugisawa et al.
Cell, 182(3), 609-624 (2020-07-09)
Gastrointestinal enterochromaffin cells regulate bone and gut homeostasis via serotonin (5-hydroxytryptamine [5-HT]) production. A recent report suggested that gut microbes regulate 5-HT levels; however, the precise underlying molecular mechanisms are unexplored. Here, we reveal that the cation channel Piezo1 in
Immacolata Andolfo et al.
American journal of hematology, 95(2), 188-197 (2019-11-19)
Dehydrated hereditary stomatocytosis (DHS), or xerocytosis, is an autosomal dominant hemolytic anemia. Most patients with DHS carry mutations in the PIEZO1 gene encoding a mechanosensitive cation channel. We here demonstrate that patients with DHS have low levels of hepcidin and
Lei Liu et al.
Frontiers in pharmacology, 12, 606245-606245 (2021-04-13)
XueShuanTong (XST) comprising therapeutically active ginsenosides, a lyophilized extract of Panax notoginseng roots, is extensively used in traditional Chinese medicine to treat ischemic heart and cerebrovascular diseases. Our recent study shows that treatment with XST inhibits shear-induced thrombosis formation but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service