Skip to Content
Merck
All Photos(1)

Key Documents

M5441

Sigma-Aldrich

Mibefradil dihydrochloride hydrate

≥98% (HPLC), powder, T-type Ca²⁺ channel blocker

Synonym(s):

(1S,2S)-2-[2[[3-(2-Benzimidazolyl)propyl]methylamino]ethyl]-6-fluoro-1,2,3,4-tetrahydro-1-isopropyl-2-naphthyl methoxyacetate dihydrochloride hydrate, Ro 40-5967 hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H38FN3O3 · 2HCl · xH2O
CAS Number:
Molecular Weight:
568.55 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Mibefradil dihydrochloride hydrate, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

H2O: soluble >20 mg/mL
absolute ethanol: insoluble

originator

Roche

SMILES string

O.Cl.Cl.COCC(=O)O[C@]3(CCN(C)CCCc1nc2ccccc2[nH]1)CCc4cc(F)ccc4[C@@H]3C(C)C

InChI

1S/C29H38FN3O3.2ClH.H2O/c1-20(2)28-23-12-11-22(30)18-21(23)13-14-29(28,36-27(34)19-35-4)15-17-33(3)16-7-10-26-31-24-8-5-6-9-25(24)32-26;;;/h5-6,8-9,11-12,18,20,28H,7,10,13-17,19H2,1-4H3,(H,31,32);2*1H;1H2/t28-,29-;;;/m0.../s1

InChI key

IZSWBBGKLWFDOC-YKXHUFBBSA-N

Application

Mibefradil has been used as a T-type calcium channel blocker in various cells. It has also been used to reduce apoptosis of retinal ganglion cells (RGC) in EphB2-Fc.

Biochem/physiol Actions

Mibefradil is a tetralol derivative and nondihydropyridine calcium channel blocker. It blocks long-type (L-type) calcium channels. It blocks mildly the Purkinje-type (P-type) calcium channels in Purkinje neurons.
Ro 40-5967 is a T-type Ca2+ channel blocker; antihypertensive.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium channels are involved in EphB/ephrinB reverse signaling-induced apoptosis in a rat chronic ocular hypertension model
Dong L, et al.
Molecular Medicine Reports, 17(2), 2465-2471 (2018)
Mibefradil block of cloned T-type calcium channels
Martin R L, et al.
Journal of Pharmacology and Experimental Therapeutics, 295(1), 302-308 (2000)
Bobbi-Jo Lowie et al.
American journal of physiology. Gastrointestinal and liver physiology, 301(5), G835-G845 (2011-08-13)
Interstitial cells of Cajal associated with the myenteric plexus (ICC-MP) are pacemaker cells of the small intestine, producing the characteristic omnipresent electrical slow waves, which orchestrate peristaltic motor activity and are associated with rhythmic intracellular calcium oscillations. Our objective was
Mibefradil inhibition of T-type calcium channels in cerebellar purkinje neurons
McDonough S I and Bean B P
Molecular Pharmacology, 54(6), 1080-1087 (1998)
Life-threatening interaction of mibefradil and beta-blockers with dihydropyridine calcium channel blockers
Mullins M E, et al.
JAMA : The Journal of the American Medical Association, 280(2), 157-158 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service