Skip to Content
Merck
All Photos(1)

Documents

G0798

Sigma-Aldrich

GSK1016790A

≥98% (HPLC), solid, TRPV4 channel agonist

Synonym(s):

GSK101, (N-((1S)-1-{[4-((2S)-2-{[(2,4-Dichlorophenyl)sulfonyl]amino}-3-hydroxypropanoyl)-1-piperazinyl]carbonyl}-3-methylbutyl)-1-benzothiophene-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32Cl2N4O6S2
CAS Number:
Molecular Weight:
655.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

GSK1016790A, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >10 mg/mL

originator

GlaxoSmithKline

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)c1cc2ccccc2s1)C(=O)N3CCN(CC3)C(=O)[C@H](CO)NS(=O)(=O)c4ccc(Cl)cc4Cl

InChI

1S/C28H32Cl2N4O6S2/c1-17(2)13-21(31-26(36)24-14-18-5-3-4-6-23(18)41-24)27(37)33-9-11-34(12-10-33)28(38)22(16-35)32-42(39,40)25-8-7-19(29)15-20(25)30/h3-8,14-15,17,21-22,32,35H,9-13,16H2,1-2H3,(H,31,36)/t21-,22-/m0/s1

InChI key

IVYQPSHHYIAUFO-VXKWHMMOSA-N

Application

GSK1016790A has been used:
  • to stimulate transient receptor potential vanilloid 4 (TRPV4)-mediated currents in Xenopus laevis oocytes
  • to stimulate cells in Ca2+ level measurement in human retinal capillary endothelial cells (HRCECs)
  • to study the role of TRPV4 channels in brain injury after intracerebral hemorrhage (ICH)
  • to trigger the opening of TRPV4 channels in bovine nucleus pulposus cells or Hanks′ Balanced Salt Solution

Biochem/physiol Actions

GSK1016790A is a novel and potent TRPV4 channel agonist. The TRPV4 (transient receptor potential vanilloid 4) member of the TRP superfamily has been implicated in numerous physiological processes. GSK1016790A elicited Ca2+ influx in mouse and human TRPV4 expressing HEK cells (EC50 values of 18 and 2.1 nM, respectively), and evoked a dose-dependent activation of TRPV4 whole-cell currents at concentrations above 1 nM. It is 300 fold more potent than 4a-PDD (Cat. No. P8014).

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TRPV4 regulates migration and tube formation of human retinal capillary endothelial cells
Wen L, et al.
BMC Ophthalmology, 18(1), 38-38 (2018)
When size matters: transient receptor potential vanilloid 4 channel as a volume-sensor rather than an osmo-sensor
Toft-Bertelsen TL, et al.
The Journal of Physiology, 595(11), 3287-3302 (2017)
TRPV4 channels stimulate Ca2+-induced Ca2+ release in mouse neurons and trigger endoplasmic reticulum stress after intracerebral hemorrhage
Shen J, et al.
Brain Research Bulletin, 146, 143-152 (2019)
Sandip M Swain et al.
The Journal of biological chemistry, 296, 100171-100171 (2020-12-11)
The ion channels Piezo1 and TRPV4 have both, independently, been implicated in high venous pressure- and fluid shear stress-induced vascular hyperpermeability in endothelial cells. However, the mechanism by which Piezo1 and TRPV4 channels execute the same function is poorly understood.
Reduced tissue osmolarity increases TRPV4 expression and pro-inflammatory cytokines in intervertebral disc cells
Walter BA, et al.
European Cells & Materials, 32, 123-123 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service