Skip to Content
Merck
All Photos(1)

Key Documents

H8502

Sigma-Aldrich

Dopamine hydrochloride

≥98% (TLC), powder, neurotransmitter

Synonym(s):

2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, 3,4-Dihydroxyphenethylamine hydrochloride, 3-Hydroxytyramine hydrochloride, 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
189.64
Beilstein:
3656720
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Product Name

Dopamine hydrochloride,

form

powder

color

light tan

mp

248-250 °C (lit.)

solubility

alcohol: 20 mg/mL
H2O: soluble

storage temp.

2-8°C

SMILES string

Cl[H].NCCc1ccc(O)c(O)c1

InChI

1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H

InChI key

CTENFNNZBMHDDG-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

General description

Dopamine (DA) works as a neurotransmitter in the central nervous system. It is a catecholamine made from the amino acid L-tyrosine. It also works as a hormone in the vesicles of the adrenal medulla, thereby controlling heart beat rate and blood pressure. The Absence of DA-containing neurons is associated with Parkinson’s disease.Dopamine is produced and released by the dopaminergic neurons in the substantia nigra that is in the central nervous system. It is involved in several activities such as movement, behaviour, learning, reward, punishment, dreaming, working, memory, mood, sleep, attention, and inhibition of prolactin production. Dopamine also acts as a precursor in the synthesis of catecholamines like epinephrine and norepinephrine. Increased levels of dopamine via the tuberoinfundibular pathway are associated with Schizophrenia, hallucinations, euphoria, and psychosis. A deficiency of dopamine and norepinephrine indicates the neurochemical basis for attention-deficit hyperactivity disorder (ADHD), a pediatric disorder.

Application

Dopamine hydrochloride has been used:
  • to study dopamine-mediated transient modulation of the physiological responses in white leg shrimp, Litopenaeus vannamei.
  • to study dopamine-mediated changes in immunity susceptibility to Lactococcus garvieae in the freshwater giant prawn, Macrobrachium rosenbergii.
  • to study the molecular link between dopamine-induced oxidative stress and mHtt (mutant Huntingtin) toxicity in relation to the activation of the autophagy pathway in an ‘in vitro’ model of parkinsonian Huntington′s Disease.
  • as a substrate of the prophenoloxidase (PPO) cascade reaction in the anti-microbial assay targeting methicillin-resistant staphylococcus aureus (MRSA) in the biofilm phase.
  • added exogenously to the culture medium to study its effect on H. contortus female worms.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Caution

Light sensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saad Latif et al.
Clinica chimica acta; international journal of clinical chemistry, 522, 114-126 (2021-08-15)
Parkinson's disease is a neurodegenerative disease caused by the death of neurons, ie, cells critical to the production of dopamine, an important neurotransmitter in the brain. Here, we present a brief review of the dopamine synthetic pathway, binding to the
Dopamine induces transient modulation of the physiological responses of whiteleg shrimp, Litopenaeus vannamei.
Chiu H-T et al.
Aquaculture (Amsterdam, Netherlands), 251, 558-566 (2006)
Natalia Del Campo et al.
Biological psychiatry, 69(12), e145-e157 (2011-05-10)
Through neuromodulatory influences over fronto-striato-cerebellar circuits, dopamine and noradrenaline play important roles in high-level executive functions often reported to be impaired in attention-deficit/hyperactivity disorder (ADHD). Medications used in the treatment of ADHD (including methylphenidate, dextroamphetamine and atomoxetine) act to increase
Dopamine depresses the immune ability and increases susceptibility to Lactococcus garvieae in the freshwater giant prawn, Macrobrachium rosenbergii.
Li JT et al.
Fish & Shellfish Immunology, 19, 269-269 (2005)
Y Luo et al.
The Journal of biological chemistry, 273(6), 3756-3764 (1998-03-07)
Dopamine (DA) is a neurotransmitter, but it also exerts a neurotoxic effect under certain pathological conditions, including age-related neurodegeneration such as Parkinson's disease. By using both the 293 cell line and primary neonatal rat postmitotic striatal neuron cultures, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service