Skip to Content
Merck
All Photos(1)

Key Documents

SML1362

Sigma-Aldrich

L755507

≥98% (HPLC)

Synonym(s):

4-[[(Hexylamino)carbonyl]amino]-N-[4-[2-[[(2S)-2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino]ethyl]phenyl]-benzenesulfonamide, L-755,507

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H40N4O6S
CAS Number:
Molecular Weight:
584.73
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC=C(OC[C@@H](O)CNCCC2=CC=C(NS(C3=CC=C(NC(NCCCCCC)=O)C=C3)(=O)=O)C=C2)C=C1

InChI

1S/C30H40N4O6S/c1-2-3-4-5-19-32-30(37)33-24-10-16-29(17-11-24)41(38,39)34-25-8-6-23(7-9-25)18-20-31-21-27(36)22-40-28-14-12-26(35)13-15-28/h6-17,27,31,34-36H,2-5,18-22H2,1H3,(H2,32,33,37)/t27-/m0/s1

InChI key

NYYJKMXNVNFOFQ-MHZLTWQESA-N

General description

L755507 is a derivative of 4-acylaminobenzenesulfonamide.

Application

L755507 has been shown to enhance CRISPR genome editing efficiency. To see other small molecule CRISPR enhancers, visit sigma.com/CRISPR-enhancers.

Biochem/physiol Actions

L755507 is a potent β3-adrenergic receptor partial agonist with an EC50 value of 0.43 nM for β3 receptors with over 440-fold selectivity for β3 compared to β1 and β2-adrenergic receptor binding. L755507 has been shown to enhance CRISPR-mediated homology-directed repair (HDR) efficiency in human induced pluripotent stem cells (iPSCs), increasing the efficiency of GFP insertion by 3-fold compared to control cells.
L755507 is a potent β3-adrenergic receptor partial agonist.
L755507 is found to stimulate the breakdown of lipids in human adipose tissues. L755507 also promotes urinary bladder relaxation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tools to study β 3-adrenoceptors.
Vrydag W and Michel M C
Naunyn-Schmiedeberg'S Archives of Pharmacology, 374(5-6), 385-398 (2007)
Dana S Hutchinson et al.
British journal of pharmacology, 135(8), 1903-1914 (2002-04-18)
1. This study characterizes the mouse beta(3a)-adrenoceptor (AR) and the splice variant of the beta(3)-AR (beta(3b)-AR) expressed in Chinese hamster ovary cells (CHO-K1). 2. Stable clones with high (approximately 1200), medium (approximately 500) or low receptor expression (approximately 100 fmol
E R Parmee et al.
Bioorganic & medicinal chemistry letters, 8(9), 1107-1112 (1999-01-01)
A study of 4-acylaminobenzenesulfonamides in a cloned human beta 3 adrenergic receptor assay resulted in the discovery of n-hexylurea, L-755,507 (22). This 0.43 nM beta 3 agonist, which is > 440-fold selective over both beta 1 and beta 2 binding
Masaaki Sato et al.
Molecular pharmacology, 74(5), 1417-1428 (2008-08-08)
This study identifies signaling pathways activated by the beta(2)-/beta(3)-adrenoceptor (AR) agonist zinterol, the selective beta(3)-AR agonist L755507, and the selective beta(3)-AR antagonist L748337 in CHO-K1 cells expressing human beta(3)-adrenoceptors. Zinterol and L755507 caused a robust concentration-dependent increase in cAMP accumulation
Ashutosh Singh et al.
The Journal of biological chemistry, 297(1), 100903-100903 (2021-06-23)
c-Myc is a transcription factor that plays a crucial role in cellular homeostasis, and its deregulation is associated with highly aggressive and chemotherapy-resistant cancers. After binding with partner MAX, the c-Myc-MAX heterodimer regulates the expression of several genes, leading to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service