Skip to Content
Merck
All Photos(4)

Key Documents

B7651

Sigma-Aldrich

Brefeldin A

from Penicillium brefeldianum, ≥99% (HPLC and TLC), powder, activator of the sphingomyelin cycle

Synonym(s):

Nectrolide, γ,4-Dihydroxy-2-(6-hydroxy-1-heptenyl)-4-cyclopentanecrotonic acid λ-lactone, Ascotoxin, BFA, Cyanein, Decumbin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O4
CAS Number:
Molecular Weight:
280.36
Beilstein:
25191
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)

biological source

Penicillium brefeldianum

Quality Level

Assay

≥99% (HPLC and TLC)

form

powder

solubility

DMSO: 10 mg/mL

antibiotic activity spectrum

neoplastics

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@H](O)\C=C\C(=O)O1

InChI

1S/C16H24O4/c1-11-5-3-2-4-6-12-9-13(17)10-14(12)15(18)7-8-16(19)20-11/h4,6-8,11-15,17-18H,2-3,5,9-10H2,1H3/b6-4+,8-7+/t11-,12+,13-,14+,15+/m0/s1

InChI key

KQNZDYYTLMIZCT-KQPMLPITSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brefeldin A (BFA) is a fungal metabolite containing a 13-membered macrocyclic lactone ring. It inhibits protein and nucleic acid synthesis. BFA blocks the exchange of GDP for GTP on adenosine ribosylation factors (ARFs), thereby hindering the binding of coat protein (β-COP) and consequently disrupts the structure and function of the Golgi apparatus. BFA is an activator of the sphingomyelin cycle. Brefeldin A-mediated apoptosis has been observed in human tumor cells. Brefeldin A has been shown to increase CRISPR-mediated homology-directed repair (HDR) efficiency. It demonstrates a diverse array of biological functions, including antitumor, antiviral, antibiotic, antimitotic, antifungal and antinematodal properties.

Application

Brefeldin A has been used to:



  • increase CRISPR genome editing efficiency.
  • facilitate the measurement of cytokine production
  • block intracellular transports in cell culture experiments

Biochem/physiol Actions

Brefeldin A (BFA) is a fungal metabolite which disrupts the structure and function of the Golgi apparatus. BFA is an activator of the sphingomyelin cycle. Brefeldin A-mediated apoptosis has been observed in human tumor cells.
Brefeldin A has been shown to increase CRISPR-mediated homology-directed repair (HDR) efficiency.
Disrupts the structure and function of the Golgi apparatus; activator of the sphingomyelin cycle.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concise Total Syntheses of (+)-Brefeldin A, Diastereomers and Analogs and Their Biological Activity
Dr. Mikhail K. Klychnikov et.al.,
Chemistry?A European Journal , 1 (2023)
Kensuke Shibata et al.
Blood, 118(3), 586-593 (2011-05-25)
Unlike conventional T cells, which are exported from the thymus as naive cells and acquire effector functions upon antigen encounter in the periphery, a subset of γδ T cells differentiates into effectors that produce IL-17 within the fetal thymus. We
Vyoma K Patel et al.
Atherosclerosis, 263, 15-23 (2017-06-02)
Atherogenesis is dependent upon monocyte influx into the vessel wall. In humans, three monocyte subsets exist, the number and function of which are significantly altered in cardiovascular disease (CVD). Whether such alterations arise in individuals with a perturbed lipid profile
Antonio Riva et al.
Frontiers in physiology, 12, 632502-632502 (2021-03-30)
Immunoregulatory checkpoint receptors (CR) contribute to the profound immunoparesis observed in alcohol-related liver disease (ALD) and in vitro neutralization of inhibitory-CRs TIM3/PD1 on anti-bacterial T-cells can rescue innate and adaptive anti-bacterial immunity. Recently described soluble-CR forms can modulate immunity in
Marco Lepore et al.
Nature communications, 5, 3866-3866 (2014-05-17)
Mucosal-associated invariant T (MAIT) cells are abundant in humans and recognize conserved bacterial antigens derived from riboflavin precursors, presented by the non-polymorphic MHC class I-like molecule MR1. Here we show that human MAIT cells are remarkably oligoclonal in both the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service