Skip to Content
Merck
All Photos(3)

Key Documents

T8552

Sigma-Aldrich

Trichostatin A

≥98% (HPLC), from Streptomyces sp.

Synonym(s):

TSA, [R-(E,E)]-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N2O3
CAS Number:
Molecular Weight:
302.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

Streptomyces sp.

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

methanol: soluble 1.90-2.10 mg/mL, clear, colorless to faint yellow or tan
DMF: soluble
DMSO: soluble
H2O: insoluble
acetone: slightly soluble
acetonitrile: soluble
benzene: slightly soluble
chloroform: slightly soluble
ethanol: soluble
ethyl acetate: slightly soluble
lower alcohols: soluble

storage temp.

−20°C

SMILES string

C[C@H](\C=C(C)\C=C\C(=O)NO)C(=O)c1ccc(cc1)N(C)C

InChI

1S/C17H22N2O3/c1-12(5-10-16(20)18-22)11-13(2)17(21)14-6-8-15(9-7-14)19(3)4/h5-11,13,22H,1-4H3,(H,18,20)/b10-5+,12-11+/t13-/m1/s1

InChI key

RTKIYFITIVXBLE-QEQCGCAPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichostatin A (TSA) is a potent and specific inhibitor of histone deacetylase (HDAC) activity. It is an antifungal antibiotic with cytostatic and differentiating properties in mammalian cell culture.

Application

Trichostatin A (TSA) has been used during the production of cloned mouse embryos. TSA has also been used as a histone deacetylase (HDAC) inhibitor in LNCaP human prostate cancer cells, as well as in MON and HeLa cells.

Biochem/physiol Actions

Trichostatin A (TSA) inhibits histone deacetylase at nanomolar concentrations; resultant histone hyperacetylation leads to chromatin relaxation and modulation of gene expression. May be involved in cell cycle progression of several cell types, inducing cell growth arrest at both G and G/M phases; may induce apoptosis. Enhances the efficacy of anticancer agents that target DNA. Trichostatin A serves as an epigenetic modifier.
Trichostatin A inhibits histone deacetylase at nanomolar concentrations; resultant histone hyperacetylation leads to chromatin relaxation and modulation of gene expression.

Features and Benefits

This compound is a featured product for Apoptosis and Gene Regulation research. Discover more featured Apoptosis and Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Preparation Note

Trichostatin A is soluble in methanol at 1.90 - 2.10 mg/ml and yields a clear, colorless to faint yellow/tan solution. It is also soluble in DMSO, DMF, acetonitrile, ethanol and lower alcohols. Trichostatin A is sparingly soluble in chloroform, ethyl acetate, acetone, and benzene. However, it is insoluble in water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Kishigami et al.
The Journal of reproduction and development, 53(6), 1207-1215 (2007-10-17)
Without using sperm, artificial oocyte activation is essential for current assisted reproductive technologies, particularly somatic cell nuclear transfer and round spermatid injection. Strontium has been widely used as an activator of oocytes especially in the mouse, by which efficient oocyte
Teena Haritwal et al.
International journal of radiation biology, 95(4), 443-451 (2018-10-12)
Ionizing radiation is known to damage male reproductive system. Current study aims to study the mitigative effects of trichostatin A on male reproductive system and accompanying metabolite changes in testicular tissue of mice. Eight-week-old male C57 Bl/6J mice were exposed
D M Vigushin et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 7(4), 971-976 (2001-04-20)
Trichostatin A (TSA), an antifungal antibiotic with cytostatic and differentiating properties in mammalian cell culture, is a potent and specific inhibitor of histone deacetylase (HDAC) activity. The purpose of this study was to evaluate the antiproliferative and HDAC inhibitory activity
Harriet E Jackson et al.
Skeletal muscle, 5(1), 2-2 (2015-02-12)
The transcription factor Sox6 has been implicated in regulating muscle fiber type-specific gene expression in mammals. In zebrafish, loss of function of the transcription factor Prdm1a results in a slow to fast-twitch fiber type transformation presaged by ectopic expression of
Cheng Liu et al.
Cancer research, 67(6), 2626-2631 (2007-03-17)
Recent evidence has accumulated that the dynamic histone methylation mediated by histone methyltransferases and demethylases plays key roles in regulation of chromatin structure and transcription. In the present study, we show that SET and MYND domain-containing protein 3 (SMYD3), a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service